Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine and canine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); Aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase
Gene Aliases: 6330403E24Rik; Ahd-1; Ahd1; ALDH5A1; D630032B01Rik; OTTMUSG00000000613; SSADH; SSDH; Ssdh1
UniProt ID: (Human) P51649, (Mouse) Q8BWF0
Entrez Gene ID: (Human) 7915, (Mouse) 214579, (Rat) 291133
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support