Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTLLINQPQ YAWLKELGLR EENEGVYNGS WGGRGEVITT YCPANNEPIA RVRQASVADY EETVKKAREA WKIWADIPAP KRGEIVRQIG DALREKIQVL GSLVSLEMGK ILVEGVGEVQ EYVDICDYAV GLSRMIGGPI LPSERSGHAL IEQWNPVGLV GIITAFNFPV AVYGWNNAIA MICGNVCLWK GAPTTSLISV AVTKIIAKVL EDNKLPGAIC SLTCGGADIG TAMAKDERVN LLSFTGSTQV GKQVGLMVQE RFGRSLLELG GNNAIIAFED ADLSLVVPSA LFAAVGTAGQ RCTTARRLFI HESIHDEVVN RLKKAYAQIR VGNPWDPNVL YGPLHTKQAV SMFLGAVEEA KKEGGTVVYG GKVMDRPGNY VEPTIVTGLG HDASIAHTET FAPILYVFKF KNEEEVFAWN NEVKQGLSSS IFTKDLGRIF RWLGPKGSDC GIVNVNIPTS GAEIGGAFGG EKHTGGGRES GSDAWKQYMR RSTCTINYSK DLPLAQGIKF Q
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family. These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular member has homology to a previously described protein from the green garden pea, the 26g pea turgor protein. It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix. Recent reports show that this protein is found both in the cytosol and the mitochondria, and the two forms likely arise from the use of alternative translation initiation sites. An additional variant encoding a different isoform has also been found for this gene. Mutations in this gene are associated with pyridoxine-dependent epilepsy. Several related pseudogenes have also been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26g turgor protein homolog; aldehyde dehydrogenase 7 family, member A1; Aldehyde dehydrogenase family 7 member A1; Alpha-AASA dehydrogenase; Alpha-aminoadipic semialdehyde dehydrogenase; Antiquitin 1; Antiquitin-1; Antiquitin1; Betaine aldehyde dehydrogenase; Delta1-piperideine-6-carboxylate dehydrogenase; P6c dehydrogenase
Gene Aliases: ALDH7A1; ATQ1; EPD; PDE
UniProt ID: (Human) P49419
Entrez Gene ID: (Human) 501
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support