Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPLLLQLAV LGAALAAAAL VLISIVAFTT ATKMPALHRH EEEKFFLNAK GQKETLPSIW DSPTKQLSVV VPSYNEEKRL PVMMDEALSY LEKRQKRDPA FTYEVIVVDD GSKDQTSKVA FKYCQKYGSD KVRVITLVKN RGKGGAIRMG IFSSRGEKIL MADADGATKF PDVEKLEKGL NDLQPWPNQM AIACGSRAHL EKESIAQRSY FRTLLMYGFH FLVWFLCVKG IRDTQCGFKL FTREAASRTF SSLHVERWAF DVELLYIAQF FKIPIAEIAV NWTEIEGSKL VPFWSWLQMG KDLLFIRLRY LTGAWRLEQT RKMN
Dolichyl-phosphate beta-glucosyltransferase (ALG5), belongs to the family of glycosyltransferases, specifically the hexosyltransferases. ALG5 is widely expressed in pancreas, placenta, liver, heart, brain, kidney, skeletal muscle, and lung. ALG5 can catalyze the chemical reaction between UDP-glucose and UDP with dolichyl phosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alg5 homolog; Alg5, S. cerevisiae, homolog of; asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferase homolog; Asparagine-linked glycosylation protein 5 homolog; dolichyl phosphate glucosyltransferase; Dolichyl-phosphate beta-glucosyltransferase; DolP-glucosyltransferase
Gene Aliases: 1500026A19Rik; 2600005J22Rik; ALG5; AV009521; AV170001; AW545564; bA421P11.2; HSPC149
UniProt ID: (Human) Q9Y673, (Mouse) Q9DB25
Entrez Gene ID: (Human) 29880, (Mouse) 66248
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support