Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLSYSRLQR KSLAVDFVVP SLFRVYARDL LLPPSSSELK AGRPEARGSL ALDCAPLLRL LGPAPGVSWT AGSPAPAEAR TLSRVLKGGS VRKLRRAKQL VLELGEEAIL EGCVGPPGEA AVGLLQFNLS ELFSWWIRQG EGRLRIRLMP EKKASEVGRE GRLSAAIRAS QPRLLFQIFG TGHSSLESPT NMPSPSPDYF TWNLTWIMKD SFPFLSHRSR YGLECSFDFP CELEYSPPLH DLRNQSWSWR RIPSEEASQM DLLDGPGAER SKEMPRGSFL LLNTSADSKH TILSPWMRSS SEHCTLAVSV HRHLQPSGRY IAQLL (43-366 aa encoded by N M_004304)
Anaplastic Lymphoma Receptor Tyrosine Kinase (ALK) belongs to the insulin receptor superfamily. It is vital for brain development. Mutations, rearrangements, and amplifications in the ALK gene have been found in tumors, including anaplastic large cell lymphoma, neurblastoma, and non-small cell lung cancer. Chromosomal rearrangement is the most common genetic alteration. The translocation creates a fusion gene consisting of the ALK gene and the nucleophosmin gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK. ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALK tyrosine kinase receptor; ALK/NPM1 fusion gene; Anaplastic lymphoma kinase; Anaplastic lymphoma kinase Ki1; Anaplastic Lymphoma Kinase p80; CD246; CD246 antigen; EC 2.7.10.1; kinase ALK; mutant anaplastic lymphoma kinase; npm-alk; TFG/ALK
Gene Aliases: ALK; CD246; NBLST3
UniProt ID: (Human) Q9UM73
Entrez Gene ID: (Human) 238
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support