Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGLYRIRVST GASLYAGSNN QVQLWLVGQH GEAALGKRLW PARGKETELK VEVPEYLGPL LFVKLRKRHL LKDDAWFCNW ISVQGPGAGD EVRFPCYRWV EGNGVLSLPE GTGRTVGEDP QGLFQKHREE ELEERRKLYR WGNWKDGLIL NMAGAKLYDL PVDERFLEDK RVDFEVSLAK GLADLAIKDS LNVLTCWKDL DDFNRIFWCG QSKLAERVRD SWKEDALFGY QFLNGANPVV LRRSAHLPAR LVFPPGMEEL QAQLEKELEG GTLFEADFSL LDGIKANVIL CSQQHLAAPL VMLKLQPDGK LLPMVIQLQL PRTGSPPPPL FLPTDPPMAW LLAKCWVRSS DFQLHELQSH LLRGHLMAEV IVVATMRCLP SIHPIFKLII PHLRYTLEIN VRARTGLVSD MGIFDQIMST GGGGHVQLLK QAGAFLTYSS FCPPDDLADR GLLGVKSSFY PQDALRLWEI IYRYVEGIVS LHYKTDVAVK DDPELQTWCR EITEIGLQGA QDRGFPVSLQ ARDQVCHFVT MCIFTCTGQH ASVHLGQLDW YSWVPNAPCT MRLPPPTTKD ATLETVMATL PNFHQASLQM SITWQLGRRQ PVMVAVGQHE EEYFSGPEPK AVLKKFREEL AALDKEIEIR NAKLDMPYEY LRPSVVENSV AI
ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12-LOX; 12/15-lipoxygenase; 15-lipooxygenase-1; 15-LOX; 15-LOX2; Arachidonate 12-lipoxygenase, leukocyte-type; Arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; Hepoxilin A3 synthase Alox15; Linoleate 13S-lipoxygenase; Polyunsaturated fatty acid lipoxygenase ALOX15
Gene Aliases: 12-LOX; 15-LOX-1; 15LOX-1; ALOX15; LOG15
UniProt ID: (Human) P16050
Entrez Gene ID: (Human) 246
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support