Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ALOX15B encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15-lipoxygenase 2; 15-LOX-2; 15-LOX-B; 15S-lipoxygenase; 8-LOX; 8S-lipoxygenase; arachidonate 15-lipoxygenase 2; Arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; Arachidonate 8S-lipoxygenase; arachidonate omega(6) lipoxygenase; Linoleate 13-lipoxygenase 15-LOb; Linoleate 9S-lipoxygenase ALOX8; Polyunsaturated fatty acid lipoxygenase ALOX15B; Polyunsaturated fatty acid lipoxygenase ALOX8
Gene Aliases: 15-LOX-2; 15-LOX-B; 8-LOX; 8S-LOX; ALOX15B; Alox8
UniProt ID: (Human) O15296, (Mouse) O35936
Entrez Gene ID: (Human) 247, (Mouse) 11688
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support