Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNNQKVVAV LLQECKQVLD QLLLEAPDVS EEDKSEDQRC RALLPSELRT LIQEAKEMKW PFVPEKWQYK QAVGPEDKTN LKDVIGAGLQ QLLASLRASI LARDCAAAAA IVFLVDRFLY GLDVSGKLLQ VAKGLHKLQP ATPIAPQVVI RQARISVNSG KLLKAEYILS SLISNNGATG TWLYRNESDK VLVQSVCIQI RGQILQKLGM WYEAAELIWA SIVGYLALPQ PDKKGLSTSL GILADIFVSM SKNDYEKFKN NPQINLSLLK EFDHHLLSAA EACKLAAAFS AYTPLFVLTA VNIRGTCLLS YSSSNDCPPE LKNLHLCEAK EAFEIGLLTK RDDEPVTG (1-347 aa encoded by BC060780)
Unlike most eukaryotic kinases, alpha kinases, such as LAK, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-kinase 1; Alpha-protein kinase 1; Chromosome 4 kinase; lymphocyte alpha-kinase; Lymphocyte alpha-protein kinase
Gene Aliases: 8430410J10Rik; ALPK1; KIAA1527; LAK
UniProt ID: (Human) Q96QP1
Entrez Gene ID: (Human) 80216, (Mouse) 71481
Molecular Function:
non-receptor serine/threonine protein kinase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support