Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: DGTSSVTEQG RYKLPTAPEA AENDYPGIQG ETRDSHQARE EFASDNLLNM DESVRETEMK LLSGESENSG MSQCWETAAD KRVGGKDLWS KRGSRKSARV RQPGMKGNP
Protein kinase that recognizes phosphorylation sites in which the surrounding peptides have an alpha-helical conformation (PubMed:10021370). Regulates cardiac development and cardiomyocyte differentiation by negatively regulating Wnt/beta-catenin signaling (PubMed:29888752). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-kinase 2; Alpha-protein kinase 2; heart alpha-kinase; Heart alpha-protein kinase; RNF18
Gene Aliases: ALPK2; HAK
UniProt ID: (Human) Q86TB3
Entrez Gene ID: (Human) 115701
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support