Product References
Role of N6-adenosine-methyltransferase subunits METTL3 and METTL14 in the biological properties of periodontal ligament cells.
Tissue & cell
Huang J,Guo C,Wang Y,Zhou Y
Thu Jun 01 00:00:00 EDT 2023
An in vivo humanized model to study homing and sequestration of Plasmodium falciparum transmission stages in the bone marrow.
Frontiers in cellular and infection microbiology
Donsante S,Siciliano G,Ciardo M,Palmisano B,Messina V,de Turris V,Farinacci G,Serafini M,Silvestrini F,Corsi A,Riminucci M,Alano P
Tue May 09 00:00:00 EDT 2023
Dihydromyricetin ameliorates osteogenic differentiation of human aortic valve interstitial cells by targeting c-KIT/interleukin-6 signaling pathway.
Frontiers in pharmacology
Zhang S,Fan L,Wang Y,Xu J,Shen Q,Xie J,Zeng Z,Zhou T
Fri Aug 26 00:00:00 EDT 2022
Dihydromyricetin ameliorates osteogenic differentiation of human aortic valve interstitial cells by targeting c-KIT/interleukin-6 signaling pathway.
Frontiers in pharmacology
Zhang S,Fan L,Wang Y,Xu J,Shen Q,Xie J,Zeng Z,Zhou T
Fri Aug 26 00:00:00 EDT 2022
Lansoprazole-induced osteoporosis via the IP3R- and SOCE-mediated calcium signaling pathways.
Molecular medicine (Cambridge, Mass.)
Cheng Z,Liu Y,Ma M,Sun S,Ma Z,Wang Y,Yu L,Qian X,Sun L,Zhang X,Liu Y,Wang Y
Sat Feb 19 00:00:00 EST 2022
Cellular communication network factor 1 interlinks autophagy and ERK signaling to promote osteogenesis of periodontal ligament stem cells.
Journal of periodontal research
Li Z,Wu Z,Xi X,Zhao F,Liu H,Liu D
Thu Dec 01 00:00:00 EST 2022
Chronic Alcohol Reduces Bone Mass Through Inhibiting Proliferation and Promoting Aging of Endothelial Cells in Type-H Vessels.
Stem cells and development
Chen A,Li X,Zhao J,Zhou J,Xie C,Chen H,Wang Q,Wang R,Miao D,Li J,Jin J
Thu Sep 01 00:00:00 EDT 2022
GsαR201C and estrogen reveal different subsets of bone marrow adiponectin expressing osteogenic cells.
Bone research
Palmisano B,Labella R,Donsante S,Remoli C,Spica E,Coletta I,Farinacci G,Dello Spedale Venti M,Saggio I,Serafini M,Robey PG,Corsi A,Riminucci M
Tue Jul 19 00:00:00 EDT 2022
Mechanical force-sensitive lncRNA SNHG8 inhibits osteogenic differentiation by regulating EZH2 in hPDLSCs.
Cellular signalling
Zhang Z,He Q,Yang S,Zhao X,Li X,Wei F
Sun May 01 00:00:00 EDT 2022
N-acetylcysteine promotes cyclic mechanical stress-induced osteogenic differentiation of periodontal ligament stem cells by down-regulating Nrf2 expression.
Journal of dental sciences
Xi X,Li ZX,Zhao Y,Liu H,Chen S,Liu DX
Fri Apr 01 00:00:00 EDT 2022
Circular RNA circSmoc1-2 regulates vascular calcification by acting as a miR-874-3p sponge in vascular smooth muscle cells.
Molecular therapy. Nucleic acids
Ryu J,Choe N,Kwon DH,Shin S,Lim YH,Yoon G,Kim JH,Kim HS,Lee IK,Ahn Y,Park WJ,Kook H,Kim YK
Tue Mar 08 00:00:00 EST 2022
Establishment and characterization of a new cell culture system for hepatitis B virus replication and infection.
Virologica Sinica
Song Y,Shou S,Guo H,Gao Z,Liu N,Yang Y,Wang F,Deng Q,Liu J,Xie Y
Mon Aug 01 00:00:00 EDT 2022
Gold nanoparticles targeting the autophagy-lysosome system to combat the inflammation-compromised osteogenic potential of periodontal ligament stem cells: From mechanism to therapy.
Biomaterials
Yin Y,Tian BM,Li X,Yu YC,Deng DK,Sun LJ,Qu HL,Wu RX,Xu XY,Sun HH,An Y,He XT,Chen FM
Thu Sep 01 00:00:00 EDT 2022
Polydopamine-Coated Poly(l-lactide) Nanofibers with Controlled Release of VEGF and BMP-2 as a Regenerative Periosteum.
ACS biomaterials science & engineering
Sun H,Dong J,Wang Y,Shen S,Shi Y,Zhang L,Zhao J,Sun X,Jiang Q
Mon Oct 11 00:00:00 EDT 2021
Cell dynamics in Hertwig's epithelial root sheath are regulated by β-catenin activity during tooth root development.
Journal of cellular physiology
Yang S,Choi H,Kim TH,Jeong JK,Liu Y,Harada H,Cho ES
Thu Jul 01 00:00:00 EDT 2021
IL-18-Mediated SLC7A5 Overexpression Enhances Osteogenic Differentiation of Human Bone Marrow Mesenchymal Stem Cells via the c-MYC Pathway.
Frontiers in cell and developmental biology
Ni F,Zhang T,Xiao W,Dong H,Gao J,Liu Y,Li J
Tue Jan 04 00:00:00 EST 2022
CDR1as regulated by hnRNPM maintains stemness of periodontal ligament stem cells via miR-7/KLF4.
Journal of cellular and molecular medicine
Gu X,Li X,Jin Y,Zhang Z,Li M,Liu D,Wei F
Sat May 01 00:00:00 EDT 2021
Nrf2 Activation Is Involved in Cyclic Mechanical Stress-Stimulated Osteogenic Differentiation in Periodontal Ligament Stem Cells via PI3K/Akt Signaling and HO1-SOD2 Interaction.
Frontiers in cell and developmental biology
Xi X,Li Z,Liu H,Chen S,Liu D
Tue Jan 25 00:00:00 EST 2022
The effects of Omarigliptin on promoting osteoblastic differentiation.
Bioengineered
Liao F,Hu X,Chen R
Wed Dec 01 00:00:00 EST 2021
Metformin coordinates osteoblast/osteoclast differentiation associated with ischemic osteonecrosis.
Aging
Park SH,Kang MA,Moon YJ,Jang KY,Kim JR
Tue Feb 11 00:00:00 EST 2020
MnTBAP inhibits bone loss in ovariectomized rats by reducing mitochondrial oxidative stress in osteoblasts.
Journal of bone and mineral metabolism
Cao X,Luo D,Li T,Huang Z,Zou W,Wang L,Lian K,Lin D
Wed Jan 01 00:00:00 EST 2020
Musashi2 promotes EGF-induced EMT in pancreatic cancer via ZEB1-ERK/MAPK signaling.
Journal of experimental & clinical cancer research : CR
Sheng W,Shi X,Lin Y,Tang J,Jia C,Cao R,Sun J,Wang G,Zhou L,Dong M
Fri Jan 17 00:00:00 EST 2020
Experimental study of expression profile and specific role of human microRNAs in regulating atrophic bone nonunion.
Medicine
Wei J,Chen H,Fu Y,Zhang B,Zhang L,Tao S,Lin F
Fri Sep 04 00:00:00 EDT 2020
Inhibition of vascular calcification by microRNA-155-5p is accompanied by the inactivation of TGF-β1/Smad2/3 signaling pathway.
Acta histochemica
Zhao F,Wu Y,Yang W,Wu D,Wang C,Zhang F
Fri May 01 00:00:00 EDT 2020
VPS4B mutation impairs the osteogenic differentiation of dental follicle cells derived from a patient with dentin dysplasia type I.
International journal of oral science
Li Q,Lu F,Chen T,Zhang K,Lu Y,Li X,Wang Y,Liu L,Tian Q,Xiong F,Chen D
Fri Jul 31 00:00:00 EDT 2020
Neuropeptide Y upregulates Runx2 and osterix and enhances osteogenesis in mouse MC3T3‑E1 cells via an autocrine mechanism.
Molecular medicine reports
Zhang B,Zhang X,Xiao J,Zhou X,Chen Y,Gao C
Sun Nov 01 00:00:00 EDT 2020
MicroRNA-21 promotes bone reconstruction in maxillary bone defects.
Journal of oral rehabilitation
Wang H,Wang H,Li X,Zhang Z,Zhao X,Wang C,Wei F
Sun Nov 01 00:00:00 EDT 2020
Effect of MicroRNA-20a on Osteogenic Differentiation of Human Adipose Tissue-Derived Stem Cells.
Cells, tissues, organs
Luo T,Yang X,Sun Y,Huang X,Zou L,Liu J
Tue Jul 07 00:00:00 EDT 2020
miR-34a promotes bone regeneration in irradiated bone defects by enhancing osteoblastic differentiation of mesenchymal stromal cells in rats.
Stem cell research & therapy
Liu H,Dong Y,Feng X,Li L,Jiao Y,Bai S,Feng Z,Yu H,Li X,Zhao Y
Tue Jun 18 00:00:00 EDT 2019
eIF2α signaling regulates autophagy of osteoblasts and the development of osteoclasts in OVX mice.
Cell death & disease
Li J,Li X,Liu D,Hamamura K,Wan Q,Na S,Yokota H,Zhang P
Wed Dec 04 00:00:00 EST 2019
Transcriptome analysis of gingival tissues of enamel-renal syndrome.
Journal of periodontal research
Wang YP,Lin HY,Zhong WL,Simmer JP,Wang SK
Sun Dec 01 00:00:00 EST 2019
Silver nanoparticles promote osteogenic differentiation of human periodontal ligament fibroblasts by regulating the RhoA-TAZ axis.
Cell biology international
Xu Y,Zheng B,He J,Cui Z,Liu Y
Thu Aug 01 00:00:00 EDT 2019
Myricetin ameliorates glucocorticoid-induced osteoporosis through the ERK signaling pathway.
Life sciences
Fan S,Gao X,Chen P,Li X
Wed Aug 15 00:00:00 EDT 2018
Myricetin ameliorates glucocorticoid-induced osteoporosis through the ERK signaling pathway.
Life sciences
Fan S,Gao X,Chen P,Li X
Wed Aug 15 00:00:00 EDT 2018
Effect of propolis on preserving human periodontal ligament cells and regulating pro-inflammatory cytokines.
Dental traumatology : official publication of International Association for Dental Traumatology
Yuan XJ,Wang YY,Shi BQ,Zhao YM
Sun May 27 00:00:00 EDT 2018
Effect of propolis on preserving human periodontal ligament cells and regulating pro-inflammatory cytokines.
Dental traumatology : official publication of International Association for Dental Traumatology
Yuan XJ,Wang YY,Shi BQ,Zhao YM
Sun May 27 00:00:00 EDT 2018
Endothelial progenitor cells improve the therapeutic effect of mesenchymal stem cell sheets on irradiated bone defect repair in a rat model.
Journal of translational medicine
Liu H,Jiao Y,Zhou W,Bai S,Feng Z,Dong Y,Liu Q,Feng X,Zhao Y
Tue May 22 00:00:00 EDT 2018
Endothelial progenitor cells improve the therapeutic effect of mesenchymal stem cell sheets on irradiated bone defect repair in a rat model.
Journal of translational medicine
Liu H,Jiao Y,Zhou W,Bai S,Feng Z,Dong Y,Liu Q,Feng X,Zhao Y
Tue May 22 00:00:00 EDT 2018
Effects of nicotine on the metabolism and gene expression profile of Sprague‑Dawley rat primary osteoblasts.
Molecular medicine reports
Liang D,Wang KJ,Tang ZQ,Liu RH,Zeng F,Cheng MY,Lian QS,Wu HK
Fri Jun 01 00:00:00 EDT 2018
The microRNA-10a/ID3/RUNX2 axis modulates the development of Ossification of Posterior Longitudinal Ligament.
Scientific reports
Xu C,Zhang H,Gu W,Wu H,Chen Y,Zhou W,Sun B,Shen X,Zhang Z,Wang Y,Liu Y,Yuan W
Fri Jun 15 00:00:00 EDT 2018
The microRNA-10a/ID3/RUNX2 axis modulates the development of Ossification of Posterior Longitudinal Ligament.
Scientific reports
Xu C,Zhang H,Gu W,Wu H,Chen Y,Zhou W,Sun B,Shen X,Zhang Z,Wang Y,Liu Y,Yuan W
Fri Jun 15 00:00:00 EDT 2018
Improvement of bone defect healing in rats via mesenchymal stem cell supernatant.
Experimental and therapeutic medicine
Zhou W,Liu Q,Xu B
Thu Feb 01 00:00:00 EST 2018
Testicular acid phosphatase induces odontoblast differentiation and mineralization.
Cell and tissue research
Choi H,Kim TH,Yun CY,Kim JW,Cho ES
Fri Apr 01 00:00:00 EDT 2016
Testicular acid phosphatase induces odontoblast differentiation and mineralization.
Cell and tissue research
Choi H,Kim TH,Yun CY,Kim JW,Cho ES
Fri Apr 01 00:00:00 EDT 2016
Disruption of Tgfbr2 in odontoblasts leads to aberrant pulp calcification.
Journal of dental research
Ahn YH,Kim TH,Choi H,Bae CH,Yang YM,Baek JA,Lee JC,Cho ES
Mon Jun 01 00:00:00 EDT 2015
Disruption of Tgfbr2 in odontoblasts leads to aberrant pulp calcification.
Journal of dental research
Ahn YH,Kim TH,Choi H,Bae CH,Yang YM,Baek JA,Lee JC,Cho ES
Mon Jun 01 00:00:00 EDT 2015
Osterix regulates tooth root formation in a site-specific manner.
Journal of dental research
Kim TH,Bae CH,Lee JC,Kim JE,Yang X,de Crombrugghe B,Cho ES
Sun Mar 01 00:00:00 EST 2015
Osterix regulates tooth root formation in a site-specific manner.
Journal of dental research
Kim TH,Bae CH,Lee JC,Kim JE,Yang X,de Crombrugghe B,Cho ES
Sun Mar 01 00:00:00 EST 2015
Magnesium ion implantation on a micro/nanostructured titanium surface promotes its bioactivity and osteogenic differentiation function.
International journal of nanomedicine
Wang G,Li J,Zhang W,Xu L,Pan H,Wen J,Wu Q,She W,Jiao T,Liu X,Jiang X
Wed Dec 10 00:00:00 EST 2014
Magnesium ion implantation on a micro/nanostructured titanium surface promotes its bioactivity and osteogenic differentiation function.
International journal of nanomedicine
Wang G,Li J,Zhang W,Xu L,Pan H,Wen J,Wu Q,She W,Jiao T,Liu X,Jiang X
Wed Dec 10 00:00:00 EST 2014
Role of Wnt/β-catenin signaling pathway in the mechanism of calcification of aortic valve.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Gu GJ,Chen T,Zhou HM,Sun KX,Li J
Sat Feb 01 00:00:00 EST 2014
Role of Wnt/β-catenin signaling pathway in the mechanism of calcification of aortic valve.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Gu GJ,Chen T,Zhou HM,Sun KX,Li J
Sat Feb 01 00:00:00 EST 2014
β-catenin is required in odontoblasts for tooth root formation.
Journal of dental research
Kim TH,Bae CH,Lee JC,Ko SO,Yang X,Jiang R,Cho ES
Fri Mar 01 00:00:00 EST 2013
New population of odontoblasts responsible for tooth root formation.
Gene expression patterns : GEP
Bae CH,Kim TH,Chu JY,Cho ES
Tue Jan 07 00:00:00 EST 2014
New population of odontoblasts responsible for tooth root formation.
Gene expression patterns : GEP
Bae CH,Kim TH,Chu JY,Cho ES
Tue Jan 07 00:00:00 EST 2014
BMP2/7 heterodimer is a stronger inducer of bone regeneration in peri-implant bone defects model than BMP2 or BMP7 homodimer.
Dental materials journal
Sun P,Wang J,Zheng Y,Fan Y,Gu Z
Tue Sep 11 00:00:00 EDT 2012
BMP2/7 heterodimer is a stronger inducer of bone regeneration in peri-implant bone defects model than BMP2 or BMP7 homodimer.
Dental materials journal
Sun P,Wang J,Zheng Y,Fan Y,Gu Z
Tue Sep 11 00:00:00 EDT 2012
Col1a1-cre mediated activation of β-catenin leads to aberrant dento-alveolar complex formation.
Anatomy & cell biology
Kim TH,Bae CH,Jang EH,Yoon CY,Bae Y,Ko SO,Taketo MM,Cho ES
Sat Sep 01 00:00:00 EDT 2012
Col1a1-cre mediated activation of β-catenin leads to aberrant dento-alveolar complex formation.
Anatomy & cell biology
Kim TH,Bae CH,Jang EH,Yoon CY,Bae Y,Ko SO,Taketo MM,Cho ES
Sat Sep 01 00:00:00 EDT 2012
Constitutive stabilization of ß-catenin in the dental mesenchyme leads to excessive dentin and cementum formation.
Biochemical and biophysical research communications
Kim TH,Lee JY,Baek JA,Lee JC,Yang X,Taketo MM,Jiang R,Cho ES
Fri Sep 09 00:00:00 EDT 2011
Constitutive stabilization of ß-catenin in the dental mesenchyme leads to excessive dentin and cementum formation.
Biochemical and biophysical research communications
Kim TH,Lee JY,Baek JA,Lee JC,Yang X,Taketo MM,Jiang R,Cho ES
Fri Sep 09 00:00:00 EDT 2011