Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody works well in WB, IHC and IF application.
Immunogen sequence: NVLYPMPAD QRRHVSINSA RWLPEPGLGL AYGTNKGDLV ICRPEALNSG VEYYWDQLNE TVFTVHSNSR SSERPGTSRA TWRTDRDMGL MNAIGLQPRN PATSVTSQGT ETLALQLQNA ETQTEREVPE PGTAASGPGE GEGSEYGASG EDALSRIQRL MAEGGMTAVV QREQSTTMAS MGGFGNNIIV SHRIHRSSQT GTEPGAAHTS SPQPSTSRGL LPEAGQLAER GLSPRTASWD QPGTPGREPT QPTLPSSSPV PIPVSLPSAE GPTVHCELTN NNHLLDGGSS RGDAAGPRGE PRNR (906-1208 aa encoded by BC045609)
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. Beclin-1, a principal regulator of autophagosome formation, is in turn regulated by Ambra1. Ambra1 associates with Beclin-1 through a region near its center as determined by yeast two-hybrid assay. Null mutations in this gene in mice resulted in embryonic lethality with severe neural tube defects associated with autophagy impairment, accumulation of ubiquitinated proteins, unbalanced cell proliferation and excessive apoptotic death. Furthermore, down-regulation of Ambra1 in cultured cells though RNA interference decreased the level of rapamycin- and nutrient starvation-induced autophagy. Multiple isoforms of Ambra1 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activating molecule in beclin-1-regulated autophagy; activating molecule in Beclin1-regulated autophagy; Activating molecule in BECN1-regulated autophagy protein 1; autophagy/beclin-1 regulator 1; DDB1 and CUL4 associated factor 3; DDB1- and CUL4-associated factor 3; ischemia related factor NYW-1; WD repeat domain 94
Gene Aliases: 2310079H06Rik; A130023A14; AA474864; AMBRA1; AV021921; D030051N19Rik; DCAF3; KIAA1736; mKIAA1736; Nyw1; WDR94
UniProt ID: (Human) Q9C0C7, (Mouse) A2AH22
Entrez Gene ID: (Human) 55626, (Rat) 59319, (Mouse) 228361
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support