Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRGEQGAAGA RVLQFTNCRI LRGGKLLRED LWVRGGRILD PEKLFFEERR VADERRDCGG RILAPGFIDV QINGGFGVDF SQATEDVGSG VALVARRILS HGVTSFCPTL VTSPPEVYHK VVPQIPVKSG GPHGAGVLGL HLEGPFISRE KRGAHPEAHL RSFEADAFQD LLATYGPLDN VRIVTLAPEL GRSHEVIRAL TARGICVSLG HSVADLRAAE DAVWSGATFI THLFNAMLPF HHRDPGIVGL LTSDRLPAGR CIFYGMIADG THTNPAALRI AHRAHPQGLV LVTDAIPALG LGNGRHTLGQ QEVEVDGLTA YVAGERPDPL GPRSQPACQV AHDPPRACPL CSQGTKTLSG SIAPMDVCVR HFLQATGCSM ESALEAASLH PAQLLGLEKS KGTLDFGADA DFVVLDDSLH VQATYISGEL VWQADAARQ
Hydrolyzes the N-glycolyl group from N-glycolylglucosamine 6-phosphate (GlcNGc-6-P) in the N-glycolylneuraminic acid (Neu5Gc) degradation pathway. Although human is not able to catalyze formation of Neu5Gc due to the inactive CMAHP enzyme, Neu5Gc is present in food and must be degraded.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amidohydrolase domain-containing protein 2; GlcNAc 6-P deacetylase; N-acetylglucosamine-6-phosphate deacetylase; putative N-acetylglucosamine-6-phosphate deacetylase
Gene Aliases: AMDHD2; CGI-14
UniProt ID: (Human) Q9Y303
Entrez Gene ID: (Human) 51005
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support