Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRDLPLTSL ALVLSALGAL LGTEALRAEE PAVGTSGLIF REDLDWPPGS PQEPLCLVAL GGDSNGSSSP LRVVGALSAY EQAFLGAVQR ARWGPRDLAT FGVCNTGDRQ AALPSLRRLG AWLRDPGGQR LVVLHLEEVT WEPTPSLRFQ EPPPGGAGPP ELALLVLYPG PGPEVTVTRA GLPGAQSLCP SRDTRYLVLA VDRPAGAWRG SGLALTLQPR GEDSRLSTAR LQALLFGDDH RCFTRMTPAL LLLPRSEPAP LPAHGQLDTV PFPPPRPSAE LEESPPSADP FLETLTRLVR ALRVPPARAS APRLALDPDA LAGFPQGLVN LSDPAALERL LDGEEPLLLL LRPTAATTGD PAPLHDPTSA PWATALARRV AAELQAAAAE LRSLPGLPPA TAPLLARLLA LCPGGPGGLG DPLRALLLLK ALQGLRVEWR GRDPRGPGRA QRSAGATAAD GPCALRELSV DLRAERSVLI PETYQANNCQ GVCGWPQSDR NPRYGNHVVL LLKMQARGAA LARPPCCVPT AYAGKLLISL SEERISAHHV PNMVATECGC R (1-560 aa encoded by BC049194)
Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); Anti-Muellerian hormone; MIS; Muellerian-inhibiting factor; Muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
Gene Aliases: AMH; MIF; MIS
UniProt ID: (Human) P03971, (Rat) P49000
Entrez Gene ID: (Human) 268, (Rat) 25378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support