Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes p130 only.
AMOT (Angiomotin) belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. AMOT plays a central role in tight junction maintenance via the complex formed with ARHGAP17 - which acts by regulating the uptake of polarity proteins at tight junctions. AMOT is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angiomotin; angiomotin p130 isoform; angiomotin p80 isoform
Gene Aliases: AMOT; CAG-2; D0Kist1; KIAA1071; RGD1564027; Sii6
UniProt ID: (Human) Q4VCS5, (Mouse) Q8VHG2
Entrez Gene ID: (Human) 154796, (Mouse) 27494, (Rat) 300289
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support