Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK-alpha-2 chain; EC 2.7.11.1; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; prkaa2; protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Aliases: 2310008I11Rik; A830082D05; AMPK; AMPK2; AMPKa2; AMPKalpha2; PRKAA; PRKAA2
UniProt ID: (Human) P54646, (Rat) Q09137, (Mouse) Q8BRK8
Entrez Gene ID: (Human) 5563, (Rat) 78975, (Mouse) 108079
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support