Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Is a promoter of calcium phosphate mineralization, playing a critical role in the formation of the compact, mineralized, aprismatic enamel surface layer during the maturation stage of amelogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amelotin; RSTI689
Gene Aliases: AMTN; UNQ689; UNQ689/PRO1329
UniProt ID: (Human) Q6UX39
Entrez Gene ID: (Human) 401138
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support