Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEKATVPVAA ATAAEGEGSP PAVAAVAGPP AAAEVGGGVG GSSRARSASS PRGMVRVCDL LLKKKPPQQQ HHKAKRNRTC RPPSSSESSS DSDNSGGGGG GGGGGGGGGG TSSNNSEEEE DDDDEEEEVS EVESFILDQD DLENPMLETA SKLLLSGTAD GADLRTVDPE TQARLEALLE AAGIGKLSTA DGKAFADPEV LRRLTSSVSC ALDEAAAALT RMRAESTANA GQSDNRSLAE ACSEGDVNAV RKLLIEGRSV NEHTEEGESL LCLACSAGYY ELAQVLLAMH ANVEDRGIKG DITPLMAAAN GGHVKIVKLL LAHKADVNAQ SSTGNTALTY ACAGGYVDVV KVLLESGASI EDHNENGHTP LMEAGSAGHV EVARLLLENG AGINTHSNEF KESALTLACY KGHLEMVRFL LEAGADQEHK TDEMHTALME ACMDGHVEVA RLLLDSGAQV NMPADSFESP LTLAACGGHV ELAALLIERG ASLEEVNDEG YTPLMEAARE GHEEMVALLL GQGANINAQT EETQETALTL ACCGGFLEVA DFLIKAGADI ELGCSTPLME AAQEGHLELV KYLLAAGANV HATTATGDTA LTYACENGHT DVADVLLQAG ADLEHESEGG RTPLMKAARA GHVCTVQFLI SKGANVNRTT ANNDHTVLSL ACAGGHLAVV ELLLAHGADP THRLKDGSTM LIEAAKGGHT SVVCYLLDYP NNLLSAPPPD VTQLTPPSHD LTDNIFPRLV C
This gene encodes a protein with ankyrin repeats, which are associated with protein-protein interactions. Studies in mice suggest that this protein is involved in liver development. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 17; Gene trap ankyrin repeat protein; Serologically defined breast cancer antigen NY-BR-16
Gene Aliases: ANKRD17; GTAR; KIAA0697; MASK2; NY-BR-16
UniProt ID: (Human) O75179
Entrez Gene ID: (Human) 26057
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support