Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLFGFGSRR GQTAQGSIDH VYTGSGYRIR DSELQKIHRA AVKGDAAEVE RCLARRSGDL DALDKQHRTA LHLACASGHV QVVTLLVNRK CQIDVCDKEN RTPLIQAVHC QEEACAVILL EHGANPNFKD IYGNTALHYA VYSESTSLAE KLLSHGAHIE ALDKDNNTPL LFAIICKKEK MVEFLLKKKA SSHAVDRLRR SALMLAVYYD SPGIVNILLK QNIDVFAQDM CGRDAEDYAI SHHLTKIQQQ ILEHKKKILK KEKSDVGSSD ESAVSIFHEL RVDSLPASDD KDLNVATKQC VPEKVSEPLP GSSHEKGNRI VNGQGEGPPA KHPSLKPSTE VEDPAVKGAV QRKNVQTLRA EQALPVASEE EQERHERSEK KQPQVKEGNN TNKSEKIQLS ENICDSTSSA AAGRLTQQRK IGKTYPQQFP KKLKEEHDRC TLKQENEEKT NVNMLYKKNR EELERKEKQY KKEVEAKQLE PTVQSLEMKS KTARNTPNRD FHNHEEMKGL MDENCILKAD IAILRQEICT MKNDNLEKEN KYLKDIKIVK ETNAALEKYI KLNEEMITET AFRYQQELND LKAENTRLNA ELLKEKESKK RLEADIESYQ SRLAAAISKH SESVKTERNL KLALERTRDV SVQVEMSSAI SKVKAENEFL TEQLSETQIK FNALKDKFCK TRDSLRKKSL ALETVQNDLS QTQQQTQEMK EMYQNAEAKV NNSTGKWNCV EERICHLQRE NAWLVQQLDD VHQKEDHKEI VTNIQRGFIE SEKKDLVLEE KSKKLMNECD HLKESLFQYE REKTGGVVSI KEDKYFQTSR KTI
Ankyrins are membrane adaptor molecules that play important roles in coupling integral membrane proteins to the spectrin-based cytoskeleton network. Mutations of ankyrin genes lead to severe genetic diseases such as fatal cardiac arrhythmias and hereditary spherocytosis. ANKRD20A (ankyrin repeat domain-containing protein 20A) is an 823 amino acid protein that contains five ANK repeats. The gene encoding ANKRD20A maps to chromosome 9, which consists of about 145 million bases and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, and familial dysautonomia are associated with chromosome 9. Also, chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain 20 family, member A1; ankyrin repeat domain 20A; Ankyrin repeat domain-containing protein 20A1
Gene Aliases: ANKRD20A; ANKRD20A1
UniProt ID: (Human) Q5TYW2
Entrez Gene ID: (Human) 84210
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support