Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
As part of the intermicrovillar adhesion complex/IMAC plays a role in epithelial brush border differentiation, controlling microvilli organization and length. Plays a role in assembly of the complex (PubMed:26812018). May play a role in cellular response to endoplasmic reticulum stress. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and SAM domain-containing protein 4B; Harmonin-interacting ankyrin repeat-containing protein; harmonin-interacting ankyrin-repeat containing protein; Harp
Gene Aliases: ANKS4B; HARP
UniProt ID: (Human) Q8N8V4
Entrez Gene ID: (Human) 257629
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support