Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene belongs to the TMEM16 family of predicted membrane proteins, that are also known as anoctamins. While little is known about the function of this gene, mutations in this gene have been associated with some cases of autosomal dominant craniocervical dystonia. Cells from individuals with a mutation in this gene exhibited abnormalities in endoplasmic reticulum-dependent calcium signaling. Studies in rat show that the rat ortholog of this protein interacts with, and modulates the activity of a sodium-activated potassium channel. Deletion of this gene caused increased pain sensitivity in the rat model system. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ano3; Anoctamin-3; Transmembrane protein 16C; transmembrane protein 16C (eight membrane-spanning domains)
Gene Aliases: AI838058; ANO3; B230324K02Rik; C11orf25; DYT23; DYT24; GENX-3947; TMEM16C
UniProt ID: (Human) Q9BYT9, (Mouse) A2AHL0
Entrez Gene ID: (Human) 63982, (Mouse) 228432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support