Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: SW620, BT-474, OVCAR-3, HepG2, Mouse brain; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein
Immunogen sequence: VGKGAAQREF HGLGD
ANT1 catalyzes the exchange of ADP and ATP across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenine nucleotide translocase-1; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; ADP ATP translocase; ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP translocase 1; ANT 1; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; Solute carrier family 25 member 4
Gene Aliases: AAC1; Anc1; ANT; ANT 1; ANT1; AU019225; MTDPS12; PEO2; PEO3; PEOA2; SLC25A4; T1
UniProt ID: (Human) P12235, (Rat) Q05962, (Mouse) P48962
Entrez Gene ID: (Human) 291, (Rat) 85333, (Mouse) 11739
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support