Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Sequence of this protein is as follows: MGRQLAGCGD AGKKASFKMS TVHEILCKLS LEGDHSTPPS AYGSVKAYTN FDAERDALNI ETAIKTKGVD EVTIVNILTN RSNAQRQDIA FAYQRRTKKE LASALKSALS GHLETVILGL LKTPAQYDAS ELKASMKGLG TDEDSLIEII CSRTNQELQE INRVYKEMYK TDLEKDIISD TSGDFRKLMV ALAKGRRAED GSVIDYELID QDARDLYDAG VKRKGTDVPK WISIMTERSV PHLQKVFDRY KSYSPYDMLE SIRKEVKGDL ENAFLNLVQC IQNKPLYFAD RLYDSMKGKG TRDKVLIRIM VSRSEVDMLK IRSEFKRKYG KSLYYYIQQD TKGDYQKALL YLCGGDD
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 33-kDa calcimedin; 33-kDa lymphocyte Ca<2+>- binding protein; 33kDa lymphocyte Ca<2+>- binding protein; 36-kDa calelectrin; Annexin A2; Annexin II; Annexin-2; Calpactin I heavy chain; calpactin I heavy polypeptide; Calpactin-1 heavy chain; chromobindin 8; Chromobindin-8; epididymis secretory protein Li 270; Lipocortin II; p36; PAP-IV; Placental anticoagulant protein IV; Protein I
Gene Aliases: ANX2; ANX2L4; ANXA2; AW215814; CAL1H; HEL-S-270; LIP2; LPC2; LPC2D; P36; PAP-IV
UniProt ID: (Human) Q9UDH8, (Mouse) P07356
Entrez Gene ID: (Human) 302, (Mouse) 12306
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support