Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aldehyde oxidase; aldehyde oxidase (female form); Aldehyde oxidase 1; aldehyde oxidase 2; Azaheterocycle hydroxylase; Azaheterocycle hydroxylase 1; Retinal oxidase
Gene Aliases: AI196512; AI255253; AO; AOH1; Aox-1; Aox-2; AOX1; Aox2; Moro; Ro
UniProt ID: (Human) Q06278, (Mouse) O54754, (Rat) Q9Z0U5
Entrez Gene ID: (Human) 316, (Mouse) 11761, (Rat) 54349
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support