Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTAKMETTF YDDALNASFL PSESGPYGYS NPKILKQSMT LNLADPVGSL KPHLRAKNSD LLTSPDVGLL KLASPELERL IIQSSNGHIT TTPTPTQFLC PKNVTDEQEG FAEGFVRALA ELHSQNTLPS VTSAAQPVNG AGMVAPAVAS VAGGSGSGGF SASLHSEPPV YANLSNFNPG ALSSGGGAPS YGAAGLAFPA QPQQQQQPPH HLPQQMPVQH PRLQALKEEP QTVPEMPGET PPLSPIDMES QERIKAERKR MRNRIAASKC RKRKLERIAR LEEKVKTLKA QNSELASTAN MLREQVAQLK QKVMNHVNSG CQLMLTQQLQ TF (1-331 aa encoded by BC002646)
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator protein 1; AH119; AP1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; enhancer-binding protein AP1; immediate early; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; JUND; p39; Proto-oncogene c-Jun; transcription factor AP-1; Transcription factor AP-1 subunit Jun; Transcription factor Jun; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Gene Aliases: AP-1; AP1; c-Jun; JUN; Junc; Rjg-9
UniProt ID: (Human) P05412, (Rat) P17325, (Mouse) P05627
Entrez Gene ID: (Human) 3725, (Rat) 24516, (Mouse) 16476
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support