Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSASAVFIL DVKGKPLISR NYKGDVAMSK IEHFMPLLVQ REEEGALAPL LSHGQVHFLW IKHSNLYLVA TTSKNANASL VYSFLYKTIE VFCEYFKELE EESIRDNFVI VYELLDELMD FGFPQTTDSK ILQEYITQQS NKLETGKSRV PPTVTNAVSW RSEGIKYKKN EVFIDVIESV NLLVNANGSV LLSEIVGTIK LKVFLSGMPE LRLGLNDRVL FELTGLSGSK NKSVELEDVK FHQCVRLSRF DNDRTISFIP PDGDFELMSY RLSTQVKPLI WIESVIEKFS HSRVEIMVKA KGQFKKQSVA NGVEISVPVP S (1-320 aa encoded by BC005021)
This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 1 (AP-1), which belongs to the adaptor complexes medium subunits family. This protein is capable of interacting with tyrosine-based sorting signals. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adaptor protein complex AP-1 mu-2 subunit; Adaptor protein complex AP-1 subunit mu-2; adaptor-related protein complex 1 mu 2 subunit; adaptor-related protein complex 1 mu-2 subunit; Adaptor-related protein complex 1 subunit mu-2; adaptor-related protein complex 1, mu 2 subunit; adaptor-related protein complex AP-1, mu 2 subunit; AP-1 complex subunit mu-2; AP-mu chain family member mu1B; clathrin assembly protein complex 1 medium chain 2; Clathrin assembly protein complex 1 mu-2 medium chain 2; clathrin coat assembly protein AP47 2; clathrin coat associated protein AP47 2; clathrin-associated adaptor medium chain mu2; golgi adaptor AP-1 47 kDa protein; Golgi adaptor HA1/AP1 adaptin mu-2 subunit; HA1 47 kDa subunit 2; Mu-adaptin 2; Mu1B-adaptin
Gene Aliases: [m]1B; AP1-mu2; AP1M2; D9Ertd818e; HSMU1B; MU-1B; MU1B; mu2; RGD1561490
UniProt ID: (Human) Q9Y6Q5, (Mouse) Q9WVP1
Entrez Gene ID: (Human) 10053, (Rat) 367038, (Mouse) 11768
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support