Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive test controls include: SKOV3, SW620, Mouse liver. The target is usually found in the following locations: Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus, Peripheral membrane protein, clathrin-coated vesicle membrane.
Immunogen sequence: FGDKMVSIQI TLNNTTDRKI ENIHIGEKKL PIGMKMHVFN PIDSLEPEGS ITVSMGIDFC DSTQTASFQL CTKDDCFNVN IQPPVGELLL PVAMSEKDFK KEQGVLTGMN ETSAVIIAAP QNFTPSVIFQ KVVNVANVGA VPSGQDNIHR FAAKTVHSGS LMLVTVELKE GSTAQLIINT EKTVIGSVLL RELKPVLSQG
AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; Adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3, beta 1 subunit; Adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; ap3; Beta-3A-adaptin; beta3A-adaptin; Clathrin assembly protein complex 3 beta-1 large chain; recombination induced mutation 2
Gene Aliases: ADTB3; ADTB3A; AP-3; AP3B1; AU015684; beta3A; C78395; HPS; HPS2; PE; pearl; rim2
UniProt ID: (Human) O00203, (Mouse) Q9Z1T1
Entrez Gene ID: (Human) 8546, (Mouse) 11774, (Rat) 309969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support