Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MDAKARNCLL QHREALEKDI KTSYIMDHMI SDGFLTISEE EKVRNEPTQQ QRAAMLIKMI LKKDNDSYVS FYNALLHEGY KDLAALLHDG IPVVSSSSGK DSVSGITSYV RTVLCEGGVP QRPVVFVTRK KLVNAIQQKL SKLKGEPGWV TIHGMAGCGK SVLAAEAVRD HSLLEGCFPG GVHWVSVGKQ DKSGLLMKLQ NLCTRLDQDE SFSQRLPLNI EEAKDRLRIL MLRKHPRSLL ILDDVWDSWV LKAFDSQCQI LLTTRDKSVT DSVMGPKYVV PVESSLGKEK GLEILSLFVN MKKADLPEQA HSIIKECKGS PLVVSLIGAL LRDFPNRWEY YLKQLQNKQF KRIRKSSSYD YEALDEAMSI SVEMLREDIK DYYTDLSILQ KDVKVPTKVL CILWDMETEE VEDILQEFVN KSLLFCDRNG KSFRYYLHDL QVDFLTEKNC SQLQDLHKKI ITQFQRYHQP HTLSPDQEDC MYWYNFLAYH MASAKMHKEL CALMFSLDWI KAKTELVGPA HLIHEFVEYR HILDEKDCAV SENFQEFLSL NGHLLGRQPF PNIVQLGLCE PETSEVYQQA KLQAKQEVDN GMLYLEWINK KNITNLSRLV VRPHTDAVYH ACFSEDGQRI ASCGADKTLQ VFKAETGEKL LEIKAHEDEV LCCAFSTDDR FIATCSVDKK VKIWNSMTGE LVHTYDEHSE QVNCCHFTNS SHHLLLATGS SDCFLKLWDL NQKECRNTMF GHTNSVNHCR FSPDDKLLAS CSADGTLKLW DATSANERKS INVKQFFLNL EDPQEDMEVI VKCCSWSADG ARIMVAAKNK IFLWNTDSRS KVADCRGHLS WVHGVMFSPD GSSFLTSSDD QTIRLWETKK VCKNSAVMLK QEVDVVFQEN EVMVLAVDHI RRLQLINGRT GQIDYLTEAQ VSCCCLSPHL QYIAFGDENG AIEILELVNN RIFQSRFQHK KTVWHIQFTA DEKTLISSSD DAEIQVWNWQ LDKCIFLRGH QETVKDFRLL KNSRLLSWSF DGTVKVWNII TGNKEKDFVC HQGTVLSCDI SHDATKFSST SADKTAKIWS FDLLLPLHEL RGHNGCVRCS AFSVDSTLLA TGDDNGEIRI WNVSNGELLH LCAPLSEEGA ATHGGWVTDL CFSPDGKMLI SAGGYIKWWN VVTGESSQTF YTNGTNLKKI HVSPDFKTYV TVDNLGILYI LQTLE
APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APAF-1; Apoptotic protease-activating factor 1; DKFZp781B1145
Gene Aliases: APAF-1; APAF1; CED4; KIAA0413
UniProt ID: (Human) Q9UJ64
Entrez Gene ID: (Human) 317
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support