Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the cytidine deaminase enzyme family. The encoded protein forms a multiple-protein editing holoenzyme with APOBEC1 complementation factor and APOBEC1 stimulating protein . This holoenzyme is involved in the editing of C-to-U nucleotide bases in apolipoprotein B and neurofibromatosis-1 mRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APO1; apolipoprotein B mRNA editing enzyme catalytic polypeptide 1; apolipoprotein B mRNA editing enzyme complex-1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1; apolipoprotein B mRNA-editing enzyme 1; Apolipoprotein B mRNA-editing enzyme catalytic subunit 1; C->U-editing enzyme APOBEC-1; HEPR; mRNA(cytosine(6666)) deaminase 1
Gene Aliases: APOBEC-1; APOBEC1; BEDP; CDAR1; HEPR
UniProt ID: (Human) P41238
Entrez Gene ID: (Human) 339
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support