Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQKEEAAV ATEAASQNGE DLENLDDPEK LKELIELPPF EIVTGERLPA NFFKFQFRNV EYSSGRNKTF LCYVVEAQGK GGQVQASRGY LEDEHAAAHA EEAFFNTILP AFDPALRYNV TWYVSSSPCA ACADRIIKTL SKTKNLRLLI LVGRLFMWEE PEIQAALKKL KEAGCKLRIM KPQDFEYVWQ NFVEQEEGES KAFQPWEDIQ ENFLYYEEKL ADILK (1-224 aa encoded by BC047767)
APOBEC2 belongs to the cytidine and deoxycytidylate deaminase family. It is probable C to U editing enzyme whose physiological substrate is not yet known. It does not display detectable apoB mRNA editing and has a low intrinsic cytidine deaminase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apolipoprotein B editing complex 2; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 2; C->U-editing enzyme APOBEC-2; mRNA(cytosine(6666)) deaminase 2; OTTHUMP00000039773; probable C->U-editing enzyme APOBEC-2
Gene Aliases: APOBEC2; ARCD1; ARP1
UniProt ID: (Human) Q9Y235, (Mouse) Q9WV35
Entrez Gene ID: (Human) 10930, (Rat) 301226, (Mouse) 11811
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support