Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNPQIRNPM KAMYPGTFYF QFKNLWEAND RDETWLCFTV EGIKRRSVVS WKTGVFRNQV DSETHCHAER CFLSWFCDDI LSPNTKYQVT WYTSWSPCPD CAGEVAEFLA RHSNVNLTIF TARLYYFQYP CYQEGLRSLS QEGVAVEIMD YEDFKYCWEN FVYNDNEPFK PWKGLKTNFR LLKRRLRESL Q (1-190 aa encoded by BC011739)
APOBEC (apolipoprotein B mRNA editing enzyme, catalytic) proteins inhibit retroviruses by deaminating cytosine residues of viral RNA and DNA. The seven APOBEC3 genes or pseudogenes are found in a cluster thought to result from gene duplication on chromosome 22. Like APOBEC3G, APOBEC3F deaminates deoxycytosine to deoxyuracil in the minus strand of HIV-1 DNA, resulting in G to A hypermutation in the plus strand of DNA. Thus, APOBEC3G and APOBEC3F provide a mechanism for innate immunity to retroviruses, and are also likely contribute to sequence variation observed in many viruses. Viral infectivity factor (Vif) imparts APOBEC3G and APOBEC3F resistance to HIV through impaired translation of their mRNA and accelerated posttranslational degradation of the APOBEC3 proteins by the 26S proteasome. Interestingly, HIV-1 Vif cannot form a complex with APOBEC3G or APOBEC3F of mouse origin as it does with the human protein, and thus mouse APOBEC3G and APOBEC3F function as a potent inhibitors of wildtype HIV-1 replication, where human APOBEC3G and APOBEC3F are only able to inhibit Vif-deficient HIV-1 replication. This implies that induction of APOBEC3G and APOBEC3F activity or a method of blocking their interaction with Vif may provide a method for therapeutic intervention.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A3C; APOBEC1-like; apolipoprotein B editing enzyme catalytic polypeptide-like 3C; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3C; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3C; DNA dC->dU-editing enzyme APOBEC-3C; MGC19485; Phorbolin I; probable DNA dC->dU-editing enzyme APOBEC-3C
Gene Aliases: A3C; APOBEC1L; APOBEC3C; ARDC2; ARDC4; ARP5; bK150C2.3; PBI
UniProt ID: (Human) Q9NRW3
Entrez Gene ID: (Human) 27350
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support