Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MKPHFRNTVE RMYRDTFSYN FYNRPILSRR NTVWLCYEVK TKGPSRPPLD AKIFRGQVYS ELKYHPEMRF FHWFSKWRKL HRDQEYEVTW YISWSPCTKC TRDMATFLAE DPKVTLTIFV ARLYYFWDPD YQEALRSLCQ KRDGPRATMK IMNYDEFQHC WSKFVYSQRE LFEPWNNLPK YYILLHIMLG EILRHSMDPP TFTFNFNNEP WVRGRHETYL CYEVERMHND TWVLLNQRRG FLCNQAPHKH GFLEGRHAEL CFLDVIPFWK LDLDQDYRVT CFTSWSPCFS CAQEMAKFIS KNKHVSLCIF TARIYDDQGR CQEGLRTLAE AGAKISIMTY SEFKHCWDTF VDHQGCPFQP WDGLDEHSQD LSGRLRAILQ NQEN
The Apoliprotein B mRNA-editing, enzyme-catalytic, polypeptide-like (APOBEC) 3 is a multi-isoform member of the cytidine deaminase family of enzymes that act on monomeric nucleoside and nucleotide substrates. Similar to TRIM5-a which targets incoming retroviral capsids, APOBEC3 plays a major role in cellular defense against retroviral infection as at least two isoforms, APOBEC3G and to a lesser extent APOBEC3F, can be incorporated HIV-1 virions and induce hypermutation in the newly synthesized viral DNA and thus destabilize the viral genome. This innate mechanism of retroviral resistance is counteracted by the HIV-1 Vif protein by inducing the ubiquitization and degradation of APOBEC3G; a single amino acid substitution (D128K) blocks APOBEC3G depletion without affecting its inhibitory activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A3G; APOBEC-related cytidine deaminase; APOBEC-related protein; APOBEC-related protein 9; apolipoprotein B editing enzyme catalytic polypeptide-like 3G; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G; apolipoprotein B mRNA-editing enzyme catalytic polypeptide 3G; ARP-9; ASGP-R2; ASGPR2; CEM-15; CEM15; CLEC4H2; Deoxycytidine deaminase; DNA dC->dU editing enzyme; DNA dC->dU-editing enzyme APOBEC-3G; FLJ12740; HBXBP; OTTHUMP00000028911; phorbolin-like protein MDS019
Gene Aliases: A3G; APOBEC3G; ARCD; ARP-9; ARP9; bK150C2.7; CEM-15; CEM15; dJ494G10.1; MDS019
UniProt ID: (Human) Q9HC16
Entrez Gene ID: (Human) 60489
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support