Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ACQPEAQEG TLSPPPKLKM SRWSLVRGRM KELLETVVNR TRDGWQWFWS PSTFRGFMQT YYDDHLRDLG PLTKAWFLES KDSLLKKTHS LCPRLVCGDK DQG (26-127 aa encoded by BC020723)
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues (5-7). The human apoC-III gene maps to chromosome 11q23 and encodes a protein that may delay catabolism of triglyceride-rich particles by inhibiting lipoprotein lipase and hepatic lipase (8). The human apoC-IV gene maps to chromosome 19q13.2 and encodes a 127 amino acid protein that is primarily expressed in the liver (9,10).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-CIV; Apolipoprotein; Apolipoprotein C-IV; Apolipoprotein C4
Gene Aliases: APO-CIV; APOC-IV; APOC4
UniProt ID: (Human) P55056
Entrez Gene ID: (Human) 346
Molecular Function:
apolipoprotein
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support