Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MPGIDKLPIE ETLEDSPQTR SLLGVFEEDA TAISNYMNQL YQAMHRIYDA QNELSAATHL TSKLLKEYEK QRFPLGGDDE VMSSTLQQFS KVIDELSSCH AVLSTQLADA MMFPITQFKE RDLKEILTLK EVFQIASNDH DAAINRYSRL SKKRENDKVK YEVTEDVYTS RKKQHQTMMH YFCALNTLQY KKKIALLEPL LGYMQAQISF FKMGSENLNE QLEEFLANIG TSVQNVRREM DSDIETMQQT IEDLEVASDP LYVPDPDPTK FPVNRNLTRK AGYLNARNKT GLVSSTWDRQ FYFTQGGNLM SQARGDVAGG LAMDIDNCSV MAVDCEDRRY CFQITSFDGK KSSILQAESK KDHEEWICTI NNISKQIYLS ENPEETAARV NQSALEAVTP SPSFQQRHES LRPAAGQSRP PTARTSSSGS LGSESTNLAA LSLDSLVAPD TPIQFDIISP VCEDQPGQAK AFGQGGRRTN PFGESGGSTK SETEDSILHQ LFIVRFLGSM EVKSDDHPDV VYETMRQILA ARAIHNIFRM TESHLLVTCD CLKLIDPQTQ VTRLTFPLPC VVLYATHQEN KRLFGFVLRT SSGRSESNLS SVCYIFESNN EGEKICDSVG LAKQIALHAE LDRRASEKQK EIERVKEKQQ KELNKQKQIE KDLEEQSRLI AASSRPNQAS SEGQFVVLSS SQSEESDLGE GGKKRESEA
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signaling and insulin signaling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AKT2 interactor; DCC-interacting protein 13-alpha; Dip13-alpha; signaling adaptor protein DIP13alpha
Gene Aliases: APPL; APPL1; DIP13A; DIP13alpha; KIAA1428; MODY14
UniProt ID: (Human) Q9UKG1
Entrez Gene ID: (Human) 26060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support