Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QQRHESLRPA AGQSRPPTAR TSSSGSLGSE STNLAALSLD SLVAPDTPIQ FDIISPVCED QPGQAKAFGQ GGRRTNPFGE SGGSTKSETE DSILHQLFIV RFLGSMEVKS DDHPDVVYET MRQILAARAI HNIFRMTESH LLVTCDCLKL IDPQTQVTRL TFPLPCVVLY ATHQENKRLF GFVLRTSSGR SESNLSSVCY IFESNNEGEK ICDSVGLAKQ IALHAELDRR ASEKQKEIER VKEKQQKELN KQKQIEKDLE EQSRLIAASS RPNQASSEGQ FVVLSSSQSE ESDLGEGGKK RESEA
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signaling and insulin signaling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein APPL like; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AKT2 interactor; DCC-interacting protein 13-alpha; DIP13 alpha; Dip13-alpha; signaling adaptor protein DIP13alpha
Gene Aliases: 2900057D21Rik; 7330406P05Rik; AI585782; APPL; APPL1; AW209077; BB022931; C88264; DIP13; DIP13A; DIP13alpha; KIAA1428; MODY14; RGD1309388
UniProt ID: (Human) Q9UKG1, (Mouse) Q8K3H0
Entrez Gene ID: (Human) 26060, (Pig) 100188978, (Rabbit) 100353097, (Rat) 290537, (Mouse) 72993
Molecular Function: scaffold/adaptor protein GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support