Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor 1; ADP-ribosylation factor 2; ADP-ribosylation factor 3; ADP-ribosylation factor 4; ADP-ribosylation factor 5; ADP-ribosylation factor 6; Liver regeneration-related protein LRRG202; pan ARF; small GTP binding protein
Gene Aliases: 5430400P17Rik; AA407803; Ac1-253; AI788669; AI854770; ARF1; ARF2; ARF3; ARF4; ARF5; ARF6; AW496366
UniProt ID: (Human) P84077, (Human) P61204, (Human) P18085, (Human) P84085, (Human) P62330, (Mouse) P84078, (Rat) P84079, (Mouse) Q8BSL7, (Rat) P84082, (Rat) P61206, (Mouse) P61205, (Rat) P61751, (Mouse) P61750, (Mouse) P84084, (Rat) P84083, (Mouse) P62331, (Rat) P62332
Entrez Gene ID: (Dog) 100856517, (Human) 375, (Dog) 486562, (Human) 377, (Dog) 607073, (Human) 378, (Dog) 475206, (Human) 381, (Human) 382, (Dog) 490679, (Mouse) 11840, (Rat) 64310, (Mouse) 11841, (Rat) 79119, (Rat) 140940, (Mouse) 11842, (Rat) 79120, (Mouse) 11843, (Mouse) 11844, (Rat) 79117, (Mouse) 11845, (Rat) 79121
Molecular Function: G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support