Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGLTISSLFS RLFGKKQMRI LMVGLDAAGK TTILYKLKLG EIVTTIPTIG FNVETVEYKN ICFTVWDVGG QDRIRPLWKH YFQNTQGLIF VVDSNDRERI QEVADELQKM LLVDELRDAV LLLFANKQDL PNAMAISEMT DKLGLQSLRN RTWYVQATCA TQGTGLYEGL DWLSNELSKR
ADP-ribosylation factor 1 is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I and class III, and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor 2; ADP-ribosylation factor 4
Gene Aliases: AA407803; ARF2; ARF4
UniProt ID: (Human) P18085, (Rat) P61751, (Mouse) P61750
Entrez Gene ID: (Human) 378, (Rat) 79120, (Mouse) 11843
Molecular Function:
G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support