Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: HeLa, HepG2, Rat brain
Immunogen sequence: YFQNTQGLIF VVDSNDRERV QESADELQKM LQEDELRDAV LLVFANKQDM PNAMPVSELT DKLGLQHLRS RTWYVQATCA TQGTGLYDGL DWLSHELSKR
ADP-ribosylation factor 5 (ARF5) is a member of the human ARF gene family. These genes encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking and as activators of phospholipase D. The gene products include 6 ARF proteins and 11 ARF-like proteins and constitute 1 family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2, and ARF3), class II (ARF4 and ARF5) and class III (ARF6). The members of each class share a common gene organization. The ARF5 gene spans approximately 3.2kb of genomic DNA and contains six exons and five introns.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor 5; EFL2; EPLG3; LERK3
Gene Aliases: ARF5
UniProt ID: (Human) P84085, (Mouse) P84084, (Rat) P84083
Entrez Gene ID: (Human) 381, (Mouse) 11844, (Rat) 79117
Molecular Function: G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support