Product References
Myricetin improves pathological changes in 3×Tg-AD mice by regulating the mitochondria-NLRP3 inflammasome-microglia channel by targeting P38 MAPK signaling pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu P,Zhou Y,Shi J,Wang F,Yang X,Zheng X,Wang Y,He Y,Xie X,Pang X
Sat Jul 01 00:00:00 EDT 2023
miR-449a ameliorates acute rejection after liver transplantation via targeting procollagen-lysine1,2-oxoglutarate5-dioxygenase 1 in macrophages.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Cao ZR,Zheng WX,Jiang YX,Chai H,Gong JH,Zhao MJ,Yan P,Liu YY,Liu XY,Huang ZT,Yang H,Peng DD,Zong KZ,Wu ZJ
Wed Mar 01 00:00:00 EST 2023
Influence of the irradiated pulmonary microenvironment on macrophage and T cell dynamics.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology
Groves AM,Misra R,Clair G,Hernady E,Olson H,Orton D,Finkelstein J,Marples B,Johnston CJ
Thu Jun 01 00:00:00 EDT 2023
BMP7 Attenuates Neuroinflammation after Spinal Cord Injury by Suppressing the Microglia Activation and Inducing Microglial Polarization Via the STAT3 Pathway.
Neurochemical research
Wei X,Huang C,Chen K,Liu S,Wang M,Yang L,Wang Y
Fri Sep 01 00:00:00 EDT 2023
BMP7 Attenuates Neuroinflammation after Spinal Cord Injury by Suppressing the Microglia Activation and Inducing Microglial Polarization Via the STAT3 Pathway.
Neurochemical research
Wei X,Huang C,Chen K,Liu S,Wang M,Yang L,Wang Y
Fri Sep 01 00:00:00 EDT 2023
The ALOX5 inhibitor Zileuton regulates tumor-associated macrophage M2 polarization by JAK/STAT and inhibits pancreatic cancer invasion and metastasis.
International immunopharmacology
Hu WM,Liu SQ,Zhu KF,Li W,Yang ZJ,Yang Q,Zhu ZC,Chang J
Tue Aug 01 00:00:00 EDT 2023
Silk Fibroin and Sericin Differentially Potentiate the Paracrine and Regenerative Functions of Stem Cells Through Multiomics Analysis.
Advanced materials (Deerfield Beach, Fla.)
Zhang Y,Sheng R,Chen J,Wang H,Zhu Y,Cao Z,Zhao X,Wang Z,Liu C,Chen Z,Zhang P,Kuang B,Zheng H,Shen C,Yao Q,Zhang W
Mon May 01 00:00:00 EDT 2023
TSAb Modulates Microglia M1 Polarization via Activation of the TSHR-Mediated NF-κB Signaling Pathway.
Neuroendocrinology
Liu Y,Yang H,Wu X,Kuang Y,Ban B,Liang Z,Li L,Huang Z,Lu D,Qin Y,Liang X,Luo Z
Fri Mar 24 00:00:00 EDT 2023
Hypoxic ASCs-derived Exosomes Attenuate Colitis by Regulating Macrophage Polarization via miR-216a-5p/HMGB1 Axis.
Inflammatory bowel diseases
Qian W,Huang L,Xu Y,Lu W,Wen W,Guo Z,Zhu W,Li Y
Mon Apr 03 00:00:00 EDT 2023
A novel transgenic mouse model expressing primate-specific nuclear choline acetyltransferase: insights into potential cholinergic vulnerability.
Scientific reports
AlQot HE,Rylett RJ
Tue Feb 21 00:00:00 EST 2023
p53 inhibits the Urea cycle and represses polyamine biosynthesis in glioma cell lines.
Metabolic brain disease
Zhao Y,Chen Y,Wei L,Ran J,Wang K,Zhu S,Liu Q
Sat Apr 01 00:00:00 EDT 2023
Wutou decoction attenuates the synovial inflammation of collagen-induced arthritis rats via regulating macrophage M1/M2 type polarization.
Journal of ethnopharmacology
Lin W,Shen P,Huang Y,Han L,Ba X,Huang Y,Yan J,Li T,Xu L,Qin K,Chen Z,Tu S
Mon Jan 30 00:00:00 EST 2023
Matrix metalloproteinase-21 promotes metastasis via increasing the recruitment and M2 polarization of macrophages in HCC.
Cancer science
Zhou J,Liu L,Hu X,Feng R,Zhao N,Zhang L,Hu W,Zhang J,Huang S,Liu L,Li W,Shan Y,Jin J
Wed Feb 01 00:00:00 EST 2023
Apigenin Alleviates Autoimmune Uveitis by Inhibiting Microglia M1 Pro-Inflammatory Polarization.
Investigative ophthalmology & visual science
Shu N,Zhang Z,Wang X,Li R,Li W,Liu X,Zhang Q,Jiang Z,Tao L,Zhang L,Hou S
Mon May 01 00:00:00 EDT 2023
Hybrid Cell Membrane-Functionalized Matrixes for Modulating Inflammatory Microenvironment and Improving Bone Defect Repair.
Advanced healthcare materials
Qiao F,Lv Y
Sat Jul 01 00:00:00 EDT 2023
Spinal cannabinoid receptor 2 activation alleviates neuropathic pain by regulating microglia and suppressing P2X7 receptor.
Frontiers in molecular neuroscience
Zhou Y,Xu Y,Yang J,Yu Z,Wang W,Yuan M,Wang Y,Bai Q,Li Z
Tue Mar 28 00:00:00 EDT 2023
Sphingosine-1-Phosphate Receptor 4 Attenuates Neutrophilic Airway Inflammation in Experimental Asthma via Repressing Proinflammatory Macrophage Activation.
International journal of biological sciences
Wang S,Tian Z,Lu Y,Huang Z,Fan Y,Li B,Zheng H,Wu X,Wang M,Zhao J,Xie J
Mon Apr 17 00:00:00 EDT 2023
Myoglobin-derived iron causes wound enlargement and impaired regeneration in pressure injuries of muscle.
eLife
Nasir NJM,Heemskerk H,Jenkins J,Hamadee NH,Bunte R,Tucker-Kellogg L
Fri Jun 02 00:00:00 EDT 2023
Danggui-Shaoyao-San Promotes Amyloid-β Clearance through Regulating Microglia Polarization via Trem2 in BV2 Cells.
Journal of integrative neuroscience
Chen G,Han M,Chen Y,Lei Y,Li M,Wang L,Wang C,Hu Y,Niu J,Yang C,Mo Y,Wang Q,Yang L,Chang X
Mon May 15 00:00:00 EDT 2023
Danggui-Shaoyao-San Promotes Amyloid-β Clearance through Regulating Microglia Polarization via Trem2 in BV2 Cells.
Journal of integrative neuroscience
Chen G,Han M,Chen Y,Lei Y,Li M,Wang L,Wang C,Hu Y,Niu J,Yang C,Mo Y,Wang Q,Yang L,Chang X
Mon May 15 00:00:00 EDT 2023
γ-Glutamylcysteine rescues mice from TNBS-driven inflammatory bowel disease through regulating macrophages polarization.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Zhou J,Yan X,Bi X,Lu S,Liu X,Yang C,Shi Y,Luo L,Yin Z
Wed Mar 01 00:00:00 EST 2023
γ-Glutamylcysteine rescues mice from TNBS-driven inflammatory bowel disease through regulating macrophages polarization.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Zhou J,Yan X,Bi X,Lu S,Liu X,Yang C,Shi Y,Luo L,Yin Z
Wed Mar 01 00:00:00 EST 2023
Baicalein ameliorates Alzheimer's disease via orchestration of CX3CR1/NF-κB pathway in a triple transgenic mouse model.
International immunopharmacology
Xie XM,Hao JJ,Shi JZ,Zhou YF,Liu PF,Wang F,Zheng XM,Yu XY,Wang CC,Yan Y,Du GH,Song JK,He YY,Pang XB
Mon May 01 00:00:00 EDT 2023
Baicalein ameliorates Alzheimer's disease via orchestration of CX3CR1/NF-κB pathway in a triple transgenic mouse model.
International immunopharmacology
Xie XM,Hao JJ,Shi JZ,Zhou YF,Liu PF,Wang F,Zheng XM,Yu XY,Wang CC,Yan Y,Du GH,Song JK,He YY,Pang XB
Mon May 01 00:00:00 EDT 2023
α-Mangostin Inhibited M1 Polarization of Macrophages/Monocytes in Antigen-Induced Arthritis Mice by Up-Regulating Silent Information Regulator 1 and Peroxisome Proliferators-Activated Receptor γ Simultaneously.
Drug design, development and therapy
Wu YJ,Zhang SS,Yin Q,Lei M,Wang QH,Chen WG,Luo TT,Zhou P,Ji CL
Fri Mar 03 00:00:00 EST 2023
Modulation of gut microbiota alleviates cerebral ischemia/reperfusion injury in rats by inhibiting M1 polarization of microglia.
Frontiers in pharmacology
Li HJ,Li DQ,Zhang YL,Ding XF,Gao HT,Zhu Y,Liu J,Zhang LX,Chen J,Chen G,Yu Y
Sun May 28 00:00:00 EDT 2023
Human placental extract regulates polarization of macrophages via IRGM/NLRP3 in allergic rhinitis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wo B,Du C,Yang Y,Qi H,Liang Z,He C,Yao F,Li X
Sat Apr 01 00:00:00 EDT 2023
Geniposide ameliorates atherosclerosis by regulating macrophage polarization via perivascular adipocyte-derived CXCL14.
Journal of ethnopharmacology
He P,Wang H,Cheng S,Hu F,Zhang L,Chen W,Xu Y,Zhang Y,Gu Y,Li Z,Jin Y,Liu X,Jia Y
Thu Oct 05 00:00:00 EDT 2023
Geniposide ameliorates atherosclerosis by regulating macrophage polarization via perivascular adipocyte-derived CXCL14.
Journal of ethnopharmacology
He P,Wang H,Cheng S,Hu F,Zhang L,Chen W,Xu Y,Zhang Y,Gu Y,Li Z,Jin Y,Liu X,Jia Y
Thu Oct 05 00:00:00 EDT 2023
Engineered exosomes derived from miR-132-overexpresssing adipose stem cells promoted diabetic wound healing and skin reconstruction.
Frontiers in bioengineering and biotechnology
Ge L,Wang K,Lin H,Tao E,Xia W,Wang F,Mao C,Feng Y
Tue Mar 21 00:00:00 EDT 2023
Castor1 overexpression regulates microglia M1/M2 polarization via inhibiting mTOR pathway.
Metabolic brain disease
Hu H,Lu X,Huang L,He Y,Liu X,Wang Y,Duan C
Wed Feb 01 00:00:00 EST 2023
Castor1 overexpression regulates microglia M1/M2 polarization via inhibiting mTOR pathway.
Metabolic brain disease
Hu H,Lu X,Huang L,He Y,Liu X,Wang Y,Duan C
Wed Feb 01 00:00:00 EST 2023
Fecal microbiota transplantation and short-chain fatty acids protected against cognitive dysfunction in a rat model of chronic cerebral hypoperfusion.
CNS neuroscience & therapeutics
Su SH,Chen M,Wu YF,Lin Q,Wang DP,Sun J,Hai J
Thu Jun 01 00:00:00 EDT 2023
Ferroptosis-dependent breast cancer cell-derived exosomes inhibit migration and invasion of breast cancer cells by suppressing M2 macrophage polarization.
PeerJ
Yi C,Wu S,Duan Q,Liu L,Li L,Luo Y,Wang A
Fri Mar 24 00:00:00 EDT 2023
Ferroptosis-dependent breast cancer cell-derived exosomes inhibit migration and invasion of breast cancer cells by suppressing M2 macrophage polarization.
PeerJ
Yi C,Wu S,Duan Q,Liu L,Li L,Luo Y,Wang A
Fri Mar 24 00:00:00 EDT 2023
Ferroptosis-dependent breast cancer cell-derived exosomes inhibit migration and invasion of breast cancer cells by suppressing M2 macrophage polarization.
PeerJ
Yi C,Wu S,Duan Q,Liu L,Li L,Luo Y,Wang A
Fri Mar 24 00:00:00 EDT 2023
Ferroptosis-dependent breast cancer cell-derived exosomes inhibit migration and invasion of breast cancer cells by suppressing M2 macrophage polarization.
PeerJ
Yi C,Wu S,Duan Q,Liu L,Li L,Luo Y,Wang A
Fri Mar 24 00:00:00 EDT 2023
Functional characterization of interleukin 4 and retinoic acid signaling crosstalk during alternative macrophage activation.
Biochimica et biophysica acta. Molecular and cell biology of lipids
Pinos I,Yu J,Pilli N,Kane MA,Amengual J
Sat Apr 01 00:00:00 EDT 2023
Porous composite hydrogels with improved MSC survival for robust epithelial sealing around implants and M2 macrophage polarization.
Acta biomaterialia
Li Y,Zhang J,Wang C,Jiang Z,Lai K,Wang Y,Yang G
Wed Feb 01 00:00:00 EST 2023
Lycium barbarum polysaccharide alleviates dextran sodium sulfate-induced inflammatory bowel disease by regulating M1/M2 macrophage polarization via the STAT1 and STAT6 pathways.
Frontiers in pharmacology
Wang J,Gao H,Xie Y,Wang P,Li Y,Zhao J,Wang C,Ma X,Wang Y,Mao Q,Xia H
Tue Aug 08 00:00:00 EDT 2023
Lycium barbarum polysaccharide alleviates dextran sodium sulfate-induced inflammatory bowel disease by regulating M1/M2 macrophage polarization via the STAT1 and STAT6 pathways.
Frontiers in pharmacology
Wang J,Gao H,Xie Y,Wang P,Li Y,Zhao J,Wang C,Ma X,Wang Y,Mao Q,Xia H
Tue Aug 08 00:00:00 EDT 2023
Activation of HCA2 regulates microglial responses to alleviate neurodegeneration in LPS-induced in vivo and in vitro models.
Journal of neuroinflammation
He D,Fu S,Ye B,Wang H,He Y,Li Z,Li J,Gao X,Liu D
Wed Mar 29 00:00:00 EDT 2023
Synergistic effects of lipopolysaccharide and rotenone on dopamine neuronal damage in rats.
CNS neuroscience & therapeutics
He JY,Li DD,Wen Q,Qin TY,Long H,Zhang SB,Zhang F
Tue Aug 01 00:00:00 EDT 2023
SGLT2i alleviates epicardial adipose tissue inflammation by modulating ketone body-glyceraldehyde-3-phosphate dehydrogenase malonylation pathway.
Journal of cardiovascular medicine (Hagerstown, Md.)
Li L,Hua C,Liu X,Wang Y,Zhao L,Zhang Y,Wang L,Su P,Yang MF,Xie B
Sat Apr 01 00:00:00 EDT 2023
SGLT2i alleviates epicardial adipose tissue inflammation by modulating ketone body-glyceraldehyde-3-phosphate dehydrogenase malonylation pathway.
Journal of cardiovascular medicine (Hagerstown, Md.)
Li L,Hua C,Liu X,Wang Y,Zhao L,Zhang Y,Wang L,Su P,Yang MF,Xie B
Sat Apr 01 00:00:00 EDT 2023
SGLT2i alleviates epicardial adipose tissue inflammation by modulating ketone body-glyceraldehyde-3-phosphate dehydrogenase malonylation pathway.
Journal of cardiovascular medicine (Hagerstown, Md.)
Li L,Hua C,Liu X,Wang Y,Zhao L,Zhang Y,Wang L,Su P,Yang MF,Xie B
Sat Apr 01 00:00:00 EDT 2023
SGLT2i alleviates epicardial adipose tissue inflammation by modulating ketone body-glyceraldehyde-3-phosphate dehydrogenase malonylation pathway.
Journal of cardiovascular medicine (Hagerstown, Md.)
Li L,Hua C,Liu X,Wang Y,Zhao L,Zhang Y,Wang L,Su P,Yang MF,Xie B
Sat Apr 01 00:00:00 EDT 2023
Ectopic endometriotic stromal cells-derived lactate induces M2 macrophage polarization via Mettl3/Trib1/ERK/STAT3 signalling pathway in endometriosis.
Immunology
Gou Y,Wang H,Wang T,Wang H,Wang B,Jiao N,Yu Y,Cao Y,Wang H,Zhang Z
Wed Mar 01 00:00:00 EST 2023
Host genetic background regulates the capacity for anti-tumor antibody-dependent phagocytosis.
bioRxiv : the preprint server for biology
Glassbrook JE,Hackett JB,Muñiz MC,Bross M,Dyson G,Movahhedin N,Ullrich A,Gibson HM
Tue May 09 00:00:00 EDT 2023
Analysis and verification of the circRNA regulatory network RNO_CIRCpedia_ 4214/RNO-miR-667-5p/Msr1 axis as a potential ceRNA promoting macrophage M2-like polarization in spinal cord injury.
BMC genomics
Cao J,Pan C,Zhang J,Chen Q,Li T,He D,Cheng X
Wed Apr 05 00:00:00 EDT 2023
Regulatory Effects of Three-Dimensional Cultured Lipopolysaccharide-Pretreated Periodontal Ligament Stem Cell-Derived Secretome on Macrophages.
International journal of molecular sciences
Su Y,Ai S,Shen Y,Cheng W,Xu C,Sui L,Zhao Y
Mon Apr 10 00:00:00 EDT 2023
Celastrol-loaded biomimetic nanodrug ameliorates APAP-induced liver injury through modulating macrophage polarization.
Journal of molecular medicine (Berlin, Germany)
Zheng J,Yang N,Wan Y,Cheng W,Zhang G,Yu S,Yang B,Liu X,Chen X,Ding X,Wu L,Yu X
Thu Jun 01 00:00:00 EDT 2023
Neuron-Derived Exosomes Promote the Recovery of Spinal Cord Injury by Modulating Nerve Cells in the Cellular Microenvironment of the Lesion Area.
Molecular neurobiology
Xu Y,Zhu ZH,Xu X,Sun HT,Zheng HM,Zhang JL,Wang HH,Fang JW,Liu YZ,Huang LL,Song ZW,Liu JB
Tue Aug 01 00:00:00 EDT 2023
Neuron-Derived Exosomes Promote the Recovery of Spinal Cord Injury by Modulating Nerve Cells in the Cellular Microenvironment of the Lesion Area.
Molecular neurobiology
Xu Y,Zhu ZH,Xu X,Sun HT,Zheng HM,Zhang JL,Wang HH,Fang JW,Liu YZ,Huang LL,Song ZW,Liu JB
Tue Aug 01 00:00:00 EDT 2023
Endothelial progenitor cell-derived exosomes promote anti-inflammatory macrophages via SOCS3/JAK2/STAT3 axis and improve the outcome of spinal cord injury.
Journal of neuroinflammation
Yuan F,Peng W,Yang Y,Xu J,Liu Y,Xie Y,Huang T,Shi C,Ding Y,Li C,Qin T,Xie S,Zhu F,Lu H,Huang J,Hu J
Fri Jun 30 00:00:00 EDT 2023
T cell-mediated targeted delivery of tadalafil regulates immunosuppression and polyamine metabolism to overcome immune checkpoint blockade resistance in hepatocellular carcinoma.
Journal for immunotherapy of cancer
Wang X,Zhang Q,Zhou J,Xiao Z,Liu J,Deng S,Hong X,Huang W,Cai M,Guo Y,Huang J,Wang Y,Lin L,Zhu K
Wed Feb 01 00:00:00 EST 2023
T cell-mediated targeted delivery of tadalafil regulates immunosuppression and polyamine metabolism to overcome immune checkpoint blockade resistance in hepatocellular carcinoma.
Journal for immunotherapy of cancer
Wang X,Zhang Q,Zhou J,Xiao Z,Liu J,Deng S,Hong X,Huang W,Cai M,Guo Y,Huang J,Wang Y,Lin L,Zhu K
Wed Feb 01 00:00:00 EST 2023
Wei-Tong-Xin exerts anti-inflammatory effects through TLR4-mediated macrophages M1/M2 polarization and affects GLP-1 secretion.
The Journal of pharmacy and pharmacology
Zhang X,Yang X,Zhang S,Wang J,Wang M,Ma T,Wan M,Lv X,Yan T,Jia Y
Fri Apr 07 00:00:00 EDT 2023
Aβ Induces Neuroinflammation and Microglial M1 Polarization via cGAS-STING-IFITM3 Signaling Pathway in BV-2 Cells.
Neurochemical research
Wu Z,Tang W,Ibrahim FEEM,Chen X,Yan H,Tao C,Wang Z,Guo Y,Fu Y,Wang Q,Ge Y
Fri Sep 01 00:00:00 EDT 2023
Aβ Induces Neuroinflammation and Microglial M1 Polarization via cGAS-STING-IFITM3 Signaling Pathway in BV-2 Cells.
Neurochemical research
Wu Z,Tang W,Ibrahim FEEM,Chen X,Yan H,Tao C,Wang Z,Guo Y,Fu Y,Wang Q,Ge Y
Fri Sep 01 00:00:00 EDT 2023
Lnc-17Rik promotes the immunosuppressive function of Myeloid-Derived suppressive cells in esophageal cancer.
Cellular immunology
Wen J,Xuan B,Gao Y,Liu Y,Wang L,He L,Meng X,Zhou T,Tao Y,Guo K,Wang Y
Wed Mar 01 00:00:00 EST 2023
Klotho enhances bone regenerative function of hPDLSCs via modulating immunoregulatory function and cell autophagy.
Journal of orthopaedic surgery and research
Niu Q,Chen H,Ou Q,Yang S,Peng Y,Xie Y,Yu L,Cheng Z,Cao Y,Wang Y
Fri Jun 02 00:00:00 EDT 2023
Klotho enhances bone regenerative function of hPDLSCs via modulating immunoregulatory function and cell autophagy.
Journal of orthopaedic surgery and research
Niu Q,Chen H,Ou Q,Yang S,Peng Y,Xie Y,Yu L,Cheng Z,Cao Y,Wang Y
Fri Jun 02 00:00:00 EDT 2023
Klotho enhances bone regenerative function of hPDLSCs via modulating immunoregulatory function and cell autophagy.
Journal of orthopaedic surgery and research
Niu Q,Chen H,Ou Q,Yang S,Peng Y,Xie Y,Yu L,Cheng Z,Cao Y,Wang Y
Fri Jun 02 00:00:00 EDT 2023
Klotho enhances bone regenerative function of hPDLSCs via modulating immunoregulatory function and cell autophagy.
Journal of orthopaedic surgery and research
Niu Q,Chen H,Ou Q,Yang S,Peng Y,Xie Y,Yu L,Cheng Z,Cao Y,Wang Y
Fri Jun 02 00:00:00 EDT 2023
Reversing inflammatory microenvironment by a single intra-articular injection of multi-stimulus responsive lipogel to relieve rheumatoid arthritis and promote joint repair.
Materials today. Bio
Tao J,Yang P,Gao M,Zhang F,Wu Y,Jiang Y,Ning Y,Li Z,Ai F
Thu Jun 01 00:00:00 EDT 2023
Human umbilical cord mesenchymal stem cell-derived exosomes promote murine skin wound healing by neutrophil and macrophage modulations revealed by single-cell RNA sequencing.
Frontiers in immunology
Liu Y,Zhang M,Liao Y,Chen H,Su D,Tao Y,Li J,Luo K,Wu L,Zhang X,Yang R
Mon Apr 03 00:00:00 EDT 2023
Three-dimensional cellular visualization in mouse apical periodontitis using combined whole-mount staining and optical tissue clearing.
Journal of oral biosciences
Tazawa K,Sasaki H
Wed Mar 01 00:00:00 EST 2023
Three-dimensional cellular visualization in mouse apical periodontitis using combined whole-mount staining and optical tissue clearing.
Journal of oral biosciences
Tazawa K,Sasaki H
Wed Mar 01 00:00:00 EST 2023
Realizing Highly Efficient Sonodynamic Bactericidal Capability through the Phonon-Electron Coupling Effect Using Two-Dimensional Catalytic Planar Defects.
Advanced materials (Deerfield Beach, Fla.)
Mao C,Jin W,Xiang Y,Zhu Y,Wu J,Liu X,Wu S,Zheng Y,Cheung KMC,Yeung KWK
Wed Mar 01 00:00:00 EST 2023
Activation of glucagon-like peptide-1 receptor in microglia exerts protective effects against sepsis-induced encephalopathy via attenuating endoplasmic reticulum stress-associated inflammation and apoptosis in a mouse model of sepsis.
Experimental neurology
Yi H,Duan Y,Song R,Zhou Y,Cui Y,Liu C,Mao Z,Hu J,Zhou F
Mon May 01 00:00:00 EDT 2023
Porcine Reproductive and Respiratory Syndrome Virus Modulates the Switch of Macrophage Polarization from M1 to M2 by Upregulating MoDC-Released sCD83.
Viruses
Gong X,Ma T,Zhang Q,Wang Y,Song C,Lai M,Zhang C,Fang X,Chen X
Fri Mar 17 00:00:00 EDT 2023
Betulinic Acid Inhibits the Stemness of Gastric Cancer Cells by Regulating the GRP78-TGF-β1 Signaling Pathway and Macrophage Polarization.
Molecules (Basel, Switzerland)
Chen JL,Tai YS,Tsai HY,Hsieh CY,Chen CL,Liu CJ,Wu DC,Huang YB,Lin MW
Sat Feb 11 00:00:00 EST 2023
Peptide-anchored neutrophil membrane-coated biomimetic nanodrug for targeted treatment of rheumatoid arthritis.
Journal of nanobiotechnology
Yang N,Li M,Wu L,Song Y,Yu S,Wan Y,Cheng W,Yang B,Mou X,Yu H,Zheng J,Li X,Yu X
Fri Jan 13 00:00:00 EST 2023
Ursolic acid promotes microglial polarization toward the M2 phenotype via PPARγ regulation of MMP2 transcription.
Neurotoxicology
Wang Y,Qiu L,Deng S,Liu F,He Z,Li M,Wang Y
Mon May 01 00:00:00 EDT 2023
Material Stiffness in Cooperation with Macrophage Paracrine Signals Determines the Tenogenic Differentiation of Mesenchymal Stem Cells.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Sheng R,Liu J,Zhang W,Luo Y,Chen Z,Chi J,Mo Q,Wang M,Sun Y,Liu C,Zhang Y,Zhu Y,Kuang B,Yan C,Liu H,Backman LJ,Chen J
Thu Jun 01 00:00:00 EDT 2023
SGLT2 inhibitor dapagliflozin alleviates intramyocardial hemorrhage and adverse ventricular remodeling via suppressing hepcidin in myocardial ischemia-reperfusion injury.
European journal of pharmacology
Chen R,Zhang Y,Zhang H,Zhou H,Tong W,Wu Y,Ma M,Chen Y
Wed Jul 05 00:00:00 EDT 2023
SGLT2 inhibitor dapagliflozin alleviates intramyocardial hemorrhage and adverse ventricular remodeling via suppressing hepcidin in myocardial ischemia-reperfusion injury.
European journal of pharmacology
Chen R,Zhang Y,Zhang H,Zhou H,Tong W,Wu Y,Ma M,Chen Y
Wed Jul 05 00:00:00 EDT 2023
Berberine mitigates intracerebral hemorrhage-induced neuroinflammation in a gut microbiota-dependent manner in mice.
Aging
Liu J,Chen H,Yu T,Fu X,Qian C,Feng X
Fri Apr 07 00:00:00 EDT 2023
MAGL regulates synovial macrophage polarization vis inhibition of mitophagy in osteoarthritic pain.
Molecular medicine reports
Gu C,Chen M,Li X,Geng D,Wang C
Thu Jun 01 00:00:00 EDT 2023
MAGL regulates synovial macrophage polarization vis inhibition of mitophagy in osteoarthritic pain.
Molecular medicine reports
Gu C,Chen M,Li X,Geng D,Wang C
Thu Jun 01 00:00:00 EDT 2023
Mollugin ameliorates murine allergic airway inflammation by inhibiting Th2 response and M2 macrophage activation.
European journal of pharmacology
Li X,Hou R,Ding H,Gao X,Wei Z,Qi T,Fang L
Fri May 05 00:00:00 EDT 2023
Tumor-secreted GRP78 induces M2 polarization of macrophages by promoting lipid catabolism.
Cellular signalling
Tian J,Zhang L,La X,Fan X,Li A,Wu C,An Y,Yan S,Dong X,Wu H,Li Z
Tue Aug 01 00:00:00 EDT 2023
Interleukin-10 genetically modified clinical-grade mesenchymal stromal cells markedly reinforced functional recovery after spinal cord injury via directing alternative activation of macrophages.
Cellular & molecular biology letters
Gao T,Huang F,Wang W,Xie Y,Wang B
Thu Mar 17 00:00:00 EDT 2022
A multifunctional drug consisting of tetracycline conjugated with odanacatib for efficient periodontitis therapy.
Frontiers in pharmacology
Li D,Zhang W,Ye W,Liu Y,Li Y,Wang Y,Shi B,Zheng X,An Y,Ma Z,Hu K,Zhou H,Xue Y
Fri Nov 18 00:00:00 EST 2022
Vagus nerve stimulation promotes the M1-to-M2 transition via inhibition of TLR4/NF-κB in microglial to rescue the reperfusion injury.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association
Zhang L,Zhang X,Liu Y,Wang S,Jia G
Thu Sep 01 00:00:00 EDT 2022
Endothelial Shp2 deficiency controls alternative activation of macrophage preventing radiation-induced lung injury through notch signaling.
iScience
Liu P,Li Y,Li M,Zhou H,Zhang H,Zhang Y,Xu J,Xu Y,Zhang J,Xia B,Cheng H,Ke Y,Zhang X
Fri Mar 18 00:00:00 EDT 2022
Myoglobin promotes macrophage polarization to M1 type and pyroptosis via the RIG-I/Caspase1/GSDMD signaling pathway in CS-AKI.
Cell death discovery
Li N,Chen J,Geng C,Wang X,Wang Y,Sun N,Wang P,Han L,Li Z,Fan H,Hou S,Gong Y
Mon Feb 28 00:00:00 EST 2022
Maresin 1 enhances osteogenic potential of mesenchymal stem cells by modulating macrophage peroxisome proliferator-activated receptor-γ-mediated inflammation resolution.
Biomaterials advances
Yao D,Zou Y,Lv Y
Sat Oct 01 00:00:00 EDT 2022
Maresin 1 enhances osteogenic potential of mesenchymal stem cells by modulating macrophage peroxisome proliferator-activated receptor-γ-mediated inflammation resolution.
Biomaterials advances
Yao D,Zou Y,Lv Y
Sat Oct 01 00:00:00 EDT 2022
One-Step Synthesis of Multifunctional Chitosan Hydrogel for Full-Thickness Wound Closure and Healing.
Advanced healthcare materials
Guo S,Yao M,Zhang D,He Y,Chang R,Ren Y,Guan F
Tue Feb 01 00:00:00 EST 2022
Maresin 1 protects against lipopolysaccharide/d-galactosamine-induced acute liver injury by inhibiting macrophage pyroptosis and inflammatory response.
Biochemical pharmacology
Yang W,Tao K,Zhang P,Chen X,Sun X,Li R
Sat Jan 01 00:00:00 EST 2022
Maresin 1 protects against lipopolysaccharide/d-galactosamine-induced acute liver injury by inhibiting macrophage pyroptosis and inflammatory response.
Biochemical pharmacology
Yang W,Tao K,Zhang P,Chen X,Sun X,Li R
Sat Jan 01 00:00:00 EST 2022
Inhalation of MSC-EVs is a noninvasive strategy for ameliorating acute lung injury.
Journal of controlled release : official journal of the Controlled Release Society
Zhao R,Wang L,Wang T,Xian P,Wang H,Long Q
Sun May 01 00:00:00 EDT 2022
C1q inhibits differentiation of oligodendrocyte progenitor cells via Wnt/β-catenin signaling activation in a cuprizone-induced mouse model of multiple sclerosis.
Experimental neurology
Gao Z,Zhang C,Feng Z,Liu Z,Yang Y,Yang K,Chen L,Yao R
Tue Feb 01 00:00:00 EST 2022
MicroRNA-146a-5p-modified human umbilical cord mesenchymal stem cells enhance protection against diabetic nephropathy in rats through facilitating M2 macrophage polarization.
Stem cell research & therapy
Zhang Y,Le X,Zheng S,Zhang K,He J,Liu M,Tu C,Rao W,Du H,Ouyang Y,Li C,Wu D
Wed Apr 27 00:00:00 EDT 2022
Wharton's jelly mesenchymal stem cell-derived small extracellular vesicles as natural nanoparticles to attenuate cartilage injury via microRNA regulation.
International journal of pharmaceutics
Chen P,Tang S,Gao H,Zhang H,Chen C,Fang Z,Peng G,Weng H,Chen A,Zhang C,Qiu Z,Li S,Chen J,Chen L,Chen X
Mon Jul 25 00:00:00 EDT 2022
Exosomes derived from bone marrow mesenchymal stem cells inhibit neuroinflammation after traumatic brain injury.
Neural regeneration research
Wen L,Wang YD,Shen DF,Zheng PD,Tu MD,You WD,Zhu YR,Wang H,Feng JF,Yang XF
Thu Dec 01 00:00:00 EST 2022
Exosomes derived from bone marrow mesenchymal stem cells inhibit neuroinflammation after traumatic brain injury.
Neural regeneration research
Wen L,Wang YD,Shen DF,Zheng PD,Tu MD,You WD,Zhu YR,Wang H,Feng JF,Yang XF
Thu Dec 01 00:00:00 EST 2022
Effects of ALA-PDT on the macrophages in wound healing and its related mechanisms in vivo and in vitro.
Photodiagnosis and photodynamic therapy
Li L,Yang Y,Yang Z,Zheng M,Luo G,He W,Yin R
Wed Jun 01 00:00:00 EDT 2022
Evaluation of an ImmunoPET Tracer for IL-12 in a Preclinical Model of Inflammatory Immune Responses.
Frontiers in immunology
Viola NT,Glassbrook JE,Kalluri JR,Hackett JB,Wicker MN,Sternberg J,Gibson HM
Thu Jun 02 00:00:00 EDT 2022
Caffeine treatment started before injury reduces hypoxic-ischemic white-matter damage in neonatal rats by regulating phenotypic microglia polarization.
Pediatric research
Yang L,Yu X,Zhang Y,Liu N,Xue X,Fu J
Thu Dec 01 00:00:00 EST 2022
Structural damage to the rat eye following long-term simulated weightlessness.
Experimental eye research
Li S,Song Q,Wu B,Kan G,Wang F,Yang J,Zhu S
Sat Oct 01 00:00:00 EDT 2022
Overexpressed DDX3x promotes abdominal aortic aneurysm formation and activates AKT in ApoE knockout mice.
Biochemical and biophysical research communications
Zhou Y,Chai H,Hu Y,Liu R,Jiang H,Fan R,Chen W,Chen X,Huang F
Sat Dec 17 00:00:00 EST 2022
Peimine ameliorates pulmonary fibrosis via the inhibition of M2-type macrophage polarization through the suppression of P38/Akt/STAT6 signals.
Bioscience reports
Cai ZH,Tian YG,Li JZ,Zhao P,Li JS,Mei X,Bai YP
Fri Oct 28 00:00:00 EDT 2022
Peimine ameliorates pulmonary fibrosis via the inhibition of M2-type macrophage polarization through the suppression of P38/Akt/STAT6 signals.
Bioscience reports
Cai ZH,Tian YG,Li JZ,Zhao P,Li JS,Mei X,Bai YP
Fri Oct 28 00:00:00 EDT 2022
Peimine ameliorates pulmonary fibrosis via the inhibition of M2-type macrophage polarization through the suppression of P38/Akt/STAT6 signals.
Bioscience reports
Cai ZH,Tian YG,Li JZ,Zhao P,Li JS,Mei X,Bai YP
Fri Oct 28 00:00:00 EDT 2022
Peimine ameliorates pulmonary fibrosis via the inhibition of M2-type macrophage polarization through the suppression of P38/Akt/STAT6 signals.
Bioscience reports
Cai ZH,Tian YG,Li JZ,Zhao P,Li JS,Mei X,Bai YP
Fri Oct 28 00:00:00 EDT 2022
An SPM-Enriched Marine Oil Supplement Shifted Microglia Polarization toward M2, Ameliorating Retinal Degeneration in rd10 Mice.
Antioxidants (Basel, Switzerland)
Olivares-González L,Velasco S,Gallego I,Esteban-Medina M,Puras G,Loucera C,Martínez-Romero A,Peña-Chilet M,Pedraz JL,Rodrigo R
Fri Dec 30 00:00:00 EST 2022
Implantation of a nerve protector embedded with human GMSC-derived Schwann-like cells accelerates regeneration of crush-injured rat sciatic nerves.
Stem cell research & therapy
Zhang Q,Burrell JC,Zeng J,Motiwala FI,Shi S,Cullen DK,Le AD
Mon Jun 20 00:00:00 EDT 2022
Protective effects of budesonide on LPS-induced podocyte injury by modulating macrophage M1/M2 polarization: Evidence from in vitro and in silico studies.
Experimental and therapeutic medicine
Zhang X,Wang G,Shen D,Feng Y,Zhang Y,Zhang C,Li Y,Liao H
Thu Sep 01 00:00:00 EDT 2022
Inhibition of IRE1 suppresses the catabolic effect of IL-1β on nucleus pulposus cell and prevents intervertebral disc degeneration in vivo.
Biochemical pharmacology
Kang H,Dong Y,Peng R,Liu H,Guo Q,Song K,Zhu M,Yu K,Wu W,Li F
Tue Mar 01 00:00:00 EST 2022
Astragali Radix-Coptis Rhizoma Herb Pair Attenuates Atherosclerosis in ApoE-/- Mice by Regulating the M1/M2 and Th1/Th2 Immune Balance and Activating the STAT6 Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Li Z,Ma D,Wang Y,Wu S,Wang L,Jiang Y,Zhang Y,Li X
Sun May 01 00:00:00 EDT 2022
Astragali Radix-Coptis Rhizoma Herb Pair Attenuates Atherosclerosis in ApoE-/- Mice by Regulating the M1/M2 and Th1/Th2 Immune Balance and Activating the STAT6 Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Li Z,Ma D,Wang Y,Wu S,Wang L,Jiang Y,Zhang Y,Li X
Sun May 01 00:00:00 EDT 2022
Injectable and reactive oxygen species-scavenging gelatin hydrogel promotes neural repair in experimental traumatic brain injury.
International journal of biological macromolecules
Zhang D,Chang R,Ren Y,He Y,Guo S,Guan F,Yao M
Mon Oct 31 00:00:00 EDT 2022
Icariin alleviates uveitis by targeting peroxiredoxin 3 to modulate retinal microglia M1/M2 phenotypic polarization.
Redox biology
Wang G,Li X,Li N,Wang X,He S,Li W,Fan W,Li R,Liu J,Hou S
Wed Jun 01 00:00:00 EDT 2022
Rutin Inhibits Ox-LDL-Mediated Macrophage Inflammation and Foam Cell Formation by Inducing Autophagy and Modulating PI3K/ATK Signaling.
Molecules (Basel, Switzerland)
Li B,Ji Y,Yi C,Wang X,Liu C,Wang C,Lu X,Xu X,Wang X
Wed Jun 29 00:00:00 EDT 2022
Effect of B7-H4 downregulation induced by Toxoplasma gondii infection on dysfunction of decidual macrophages contributes to adverse pregnancy outcomes.
Parasites & vectors
Cui L,Wang Y,Ren L,Li Z,Jiang Y,Wang C,Liu X,Ren Y,Hu X
Tue Dec 13 00:00:00 EST 2022
Urea transporter B downregulates polyamines levels in melanoma B16 cells via p53 activation.
Biochimica et biophysica acta. Molecular cell research
Li J,Sun Y,Yan R,Wu X,Zou H,Meng Y
Sun May 01 00:00:00 EDT 2022
In Vitro Immunomodulatory Effects of Inonotus obliquus Extracts on Resting M0 Macrophages and LPS-Induced M1 Macrophages.
Evidence-based complementary and alternative medicine : eCAM
Shen D,Feng Y,Zhang X,Liu J,Gong L,Liao H,Li R
Sat Jul 16 00:00:00 EDT 2022
Mechanical stress protects against chondrocyte pyroptosis through lipoxin A4 via synovial macrophage M2 subtype polarization in an osteoarthritis model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Shen P,Jia S,Wang Y,Zhou X,Zhang D,Jin Z,Wang Z,Liu D,Bai L,Yang Y
Thu Sep 01 00:00:00 EDT 2022
Progranulin deficiency promotes persistent neuroinflammation and causes regional pathology in the hippocampus following traumatic brain injury.
Glia
Zheng X,Mi T,Wang R,Zhang Z,Li W,Zhao J,Yang P,Xia H,Mao Q
Fri Jul 01 00:00:00 EDT 2022
Progranulin deficiency promotes persistent neuroinflammation and causes regional pathology in the hippocampus following traumatic brain injury.
Glia
Zheng X,Mi T,Wang R,Zhang Z,Li W,Zhao J,Yang P,Xia H,Mao Q
Fri Jul 01 00:00:00 EDT 2022
GABA regulates IL-1β production in macrophages.
Cell reports
Fu J,Han Z,Wu Z,Xia Y,Yang G,Yin Y,Ren W
Tue Dec 06 00:00:00 EST 2022
Prognostic Value and Biological Function of Galectins in Malignant Glioma.
Frontiers in oncology
Zhu H,Liu D,Cheng L,Liu J,Wang G,Li H,Zhang Y,Mi H,Zhang S,Shu K,Yu X
Sat Jul 16 00:00:00 EDT 2022
Identification of the novel prognostic biomarker, MLLT11, reveals its relationship with immune checkpoint markers in glioma.
Frontiers in oncology
Chen L,Xiong Z,Zhao H,Teng C,Liu H,Huang Q,Wanggou S,Li X
Tue Aug 30 00:00:00 EDT 2022
miR-182 targeting reprograms tumor-associated macrophages and limits breast cancer progression.
Proceedings of the National Academy of Sciences of the United States of America
Ma C,He D,Tian P,Wang Y,He Y,Wu Q,Jia Z,Zhang X,Zhang P,Ying H,Jin ZB,Hu G
Tue Feb 08 00:00:00 EST 2022
Grape Seed Proanthocyanidins Exert a Neuroprotective Effect by Regulating Microglial M1/M2 Polarisation in Rats with Spinal Cord Injury.
Mediators of inflammation
Liu WZ,Ma ZJ,Kang JH,Lin AX,Wang ZH,Chen HW,Guo XD,He XG,Kang XW
Tue Aug 16 00:00:00 EDT 2022
Identification and validation of a 17-gene signature to improve the survival prediction of gliomas.
Frontiers in immunology
Tong S,Xia M,Xu Y,Sun Q,Ye L,Cai J,Ye Z,Tian D
Tue Oct 18 00:00:00 EDT 2022
Identification and validation of a 17-gene signature to improve the survival prediction of gliomas.
Frontiers in immunology
Tong S,Xia M,Xu Y,Sun Q,Ye L,Cai J,Ye Z,Tian D
Tue Oct 18 00:00:00 EDT 2022
The Clostridium Metabolite P-Cresol Sulfate Relieves Inflammation of Primary Biliary Cholangitis by Regulating Kupffer Cells.
Cells
Fu HY,Xu JM,Ai X,Dang FT,Tan X,Yu HY,Feng J,Yang WX,Ma HT,Tu RF,Gupta AK,Manandhar LK,Bao WM,Tang YM
Sat Nov 26 00:00:00 EST 2022
Novel recombinant protein flagellin A N/C attenuates experimental autoimmune encephalomyelitis by suppressing the ROS/NF-κB/NLRP3 signaling pathway.
Frontiers in pharmacology
Li L,Deng S,Liu M,Yang M,Li J,Liu T,Zhang T,Zhao Y,He M,Wu D,Xu Y
Fri Dec 02 00:00:00 EST 2022
Injectable hyaluronic acid hydrogel loaded with BMSC and NGF for traumatic brain injury treatment.
Materials today. Bio
Wang L,Zhang D,Ren Y,Guo S,Li J,Ma S,Yao M,Guan F
Sat Jan 01 00:00:00 EST 2022
Schisandra chinensis Lignans Exert Antidepressant Effects by Promoting BV2 Microglia Polarization toward the M2 Phenotype through the Activation of the Cannabinoid Receptor Type-2-Signal Transducer and Activator of Transcription 6 Pathway.
Journal of agricultural and food chemistry
Wang J,Zhang G,Yang Y,Zhang X,Shi K,Zhang X,Yan T,Jia Y
Wed Nov 09 00:00:00 EST 2022
Gut microbiota-derived butyrate regulates gut mucus barrier repair by activating the macrophage/WNT/ERK signaling pathway.
Clinical science (London, England : 1979)
Liang L,Liu L,Zhou W,Yang C,Mai G,Li H,Chen Y
Fri Feb 25 00:00:00 EST 2022
Gut microbiota-derived butyrate regulates gut mucus barrier repair by activating the macrophage/WNT/ERK signaling pathway.
Clinical science (London, England : 1979)
Liang L,Liu L,Zhou W,Yang C,Mai G,Li H,Chen Y
Fri Feb 25 00:00:00 EST 2022
OIT3 mediates macrophage polarization and facilitates hepatocellular carcinoma progression.
Cancer immunology, immunotherapy : CII
Yang S,Zhang J,Xu Y,Wang J,Zhao H,Lei J,Zhou Y,Chen Y,Wu L,Zhou M,Zheng L,Ji X,Li Y
Tue Nov 01 00:00:00 EDT 2022
Inhibition of Mer exacerbates early brain injury by regulating microglia/macrophage phenotype after subarachnoid hemorrhage in mice.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association
Tan X,Zheng Y,Zeng H,Peng Y,Yu X,Cao S
Thu Sep 01 00:00:00 EDT 2022
A cell-free difunctional demineralized bone matrix scaffold enhances the recruitment and osteogenesis of mesenchymal stem cells by promoting inflammation resolution.
Biomaterials advances
Yao D,Lv Y
Mon Aug 01 00:00:00 EDT 2022
A cell-free difunctional demineralized bone matrix scaffold enhances the recruitment and osteogenesis of mesenchymal stem cells by promoting inflammation resolution.
Biomaterials advances
Yao D,Lv Y
Mon Aug 01 00:00:00 EDT 2022
Mesenchymal Stem Cells Improve Cognitive Impairment and Reduce Aβ Deposition via Promoting AQP4 Polarity and Relieving Neuroinflammation in Rats With Chronic Hypertension-Induced Cerebral Small-Vessel Disease.
Frontiers in aging neuroscience
Liu XL,Ouyang FB,Hu LT,Sun P,Yang J,Sun YJ,Liao MS,Lan LF,Pei Z,Fan YH
Sat Jul 16 00:00:00 EDT 2022
Notoginsenoside R1-loaded mesoporous silica nanoparticles targeting the site of injury through inflammatory cells improves heart repair after myocardial infarction.
Redox biology
Li H,Zhu J,Xu YW,Mou FF,Shan XL,Wang QL,Liu BN,Ning K,Liu JJ,Wang YC,Mi JX,Wei X,Shao SJ,Cui GH,Lu R,Guo HD
Mon Aug 01 00:00:00 EDT 2022
Epithelial‑derived exosomes promote M2 macrophage polarization via Notch2/SOCS1 during mechanical ventilation.
International journal of molecular medicine
Wang Y,Xie W,Feng Y,Xu Z,He Y,Xiong Y,Chen L,Li X,Liu J,Liu G,Wu Q
Fri Jul 01 00:00:00 EDT 2022
Biomimetic manganese-eumelanin nanocomposites for combined hyperthermia-immunotherapy against prostate cancer.
Journal of nanobiotechnology
Liu Y,Shang W,Liu H,Hui H,Wu J,Zhang W,Gao P,Guo K,Guo Y,Tian J
Mon Jan 24 00:00:00 EST 2022
Macrophage Extracellular Traps Exacerbate Secondary Spinal Cord Injury by Modulating Macrophage/Microglia Polarization via LL37/P2X7R/NF-κB Signaling Pathway.
Oxidative medicine and cellular longevity
Zhang C,Guo D,Qiao H,Li J,Li J,Yang Y,Chang S,Li F,Wang D,Li H,He X,Wang F
Tue Dec 06 00:00:00 EST 2022
TRIM21 aggravates cardiac injury after myocardial infarction by promoting M1 macrophage polarization.
Frontiers in immunology
Li Z,Liu X,Zhang X,Zhang W,Gong M,Qin X,Luo J,Fang Y,Liu B,Wei Y
Tue Nov 29 00:00:00 EST 2022
TRIM21 aggravates cardiac injury after myocardial infarction by promoting M1 macrophage polarization.
Frontiers in immunology
Li Z,Liu X,Zhang X,Zhang W,Gong M,Qin X,Luo J,Fang Y,Liu B,Wei Y
Tue Nov 29 00:00:00 EST 2022
CXCR3 antagonist AMG487 ameliorates experimental autoimmune prostatitis by diminishing Th1 cell differentiation and inhibiting macrophage M1 phenotypic activation.
The Prostate
Hua X,Zhang J,Ge S,Liu H,Du H,Niu Q,Chen X,Yang C,Zhang L,Liang C
Thu Sep 01 00:00:00 EDT 2022
Pseudomonas aeruginosa PcrV Enhances the Nitric Oxide-Mediated Tumoricidal Activity of Tumor-Associated Macrophages via a TLR4/PI3K/AKT/mTOR-Glycolysis-Nitric Oxide Circuit.
Frontiers in oncology
Yu H,Bai Y,Qiu J,He X,Xiong J,Dai Q,Wang X,Li Y,Sheng H,Xin R,Jiang L,Li Q,Li D,Zhang H,Zhang L,Chen Q,Peng J,Hu X,Zhang K
Wed Dec 15 00:00:00 EST 2021
CSF1/CSF1R-mediated Crosstalk Between Choroidal Vascular Endothelial Cells and Macrophages Promotes Choroidal Neovascularization.
Investigative ophthalmology & visual science
Zhou Y,Zeng J,Tu Y,Li L,Du S,Zhu L,Cang X,Lu J,Zhu M,Liu X
Mon Mar 01 00:00:00 EST 2021
The Effect of miR-520b on Macrophage Polarization and T Cell Immunity by Targeting PTEN in Breast Cancer.
Journal of oncology
Zhu Q,Yuan J,He Y,Hu Y
Wed Apr 27 00:00:00 EDT 2022
Sodium Lactate Accelerates M2 Macrophage Polarization and Improves Cardiac Function after Myocardial Infarction in Mice.
Cardiovascular therapeutics
Zhang J,Huang F,Chen L,Li G,Lei W,Zhao J,Liao Y,Li Y,Li C,Chen M
Wed Jul 21 00:00:00 EDT 2021
Sodium Lactate Accelerates M2 Macrophage Polarization and Improves Cardiac Function after Myocardial Infarction in Mice.
Cardiovascular therapeutics
Zhang J,Huang F,Chen L,Li G,Lei W,Zhao J,Liao Y,Li Y,Li C,Chen M
Wed Jul 21 00:00:00 EDT 2021
Abdominal paracentesis drainage attenuates intestinal mucosal barrier damage through macrophage polarization in severe acute pancreatitis.
Experimental biology and medicine (Maywood, N.J.)
Yuan X,Luo C,Wu J,Li W,Guo X,Li S,Wang B,Sun H,Tang L
Wed Sep 01 00:00:00 EDT 2021
Abdominal paracentesis drainage attenuates intestinal mucosal barrier damage through macrophage polarization in severe acute pancreatitis.
Experimental biology and medicine (Maywood, N.J.)
Yuan X,Luo C,Wu J,Li W,Guo X,Li S,Wang B,Sun H,Tang L
Wed Sep 01 00:00:00 EDT 2021
PLXNA2 knockdown promotes M2 microglia polarization through mTOR/STAT3 signaling to improve functional recovery in rats after cerebral ischemia/reperfusion injury.
Experimental neurology
Li S,Hua X,Zheng M,Wu J,Ma Z,Xing X,Ma J,Zhang J,Shan C,Xu J
Wed Dec 01 00:00:00 EST 2021
Inhibition of Sphingosine-1-Phosphate Receptor 2 Prevents Thoracic Aortic Dissection and Rupture.
Frontiers in cardiovascular medicine
Pan G,Liao M,Dai Y,Li Y,Yan X,Mai W,Liu J,Liao Y,Qiu Z,Zhou Z
Tue Jan 04 00:00:00 EST 2022
Caloric restriction ameliorates high-fat diet induced cognitive deficits through attenuating neuroinflammation via the TREM2-PI3K/AKT signaling pathway.
Food & function
Wang R,Zhou Z,Wang D,Zhao Q,Zhang C,Liu C,Zhao H,Yuan C,Yuan D,Wang T
Wed Jul 21 00:00:00 EDT 2021
The glycolysis inhibitor 2-deoxyglucose ameliorates adjuvant-induced arthritis by regulating macrophage polarization in an AMPK-dependent manner.
Molecular immunology
Cai W,Cheng J,Zong S,Yu Y,Wang Y,Song Y,He R,Yuan S,Chen T,Hu M,Pan Y,Ma R,Liu H,Wei F
Wed Dec 01 00:00:00 EST 2021
The glycolysis inhibitor 2-deoxyglucose ameliorates adjuvant-induced arthritis by regulating macrophage polarization in an AMPK-dependent manner.
Molecular immunology
Cai W,Cheng J,Zong S,Yu Y,Wang Y,Song Y,He R,Yuan S,Chen T,Hu M,Pan Y,Ma R,Liu H,Wei F
Wed Dec 01 00:00:00 EST 2021
A systematic CRISPR screen reveals an IL-20/IL20RA-mediated immune crosstalk to prevent the ovarian cancer metastasis.
eLife
Li J,Qin X,Shi J,Wang X,Li T,Xu M,Chen X,Zhao Y,Han J,Piao Y,Zhang W,Qu P,Wang L,Xiang R,Shi Y
Fri Jun 11 00:00:00 EDT 2021
Inhibition of Dectin-1 Ameliorates Neuroinflammation by Regulating Microglia/Macrophage Phenotype After Intracerebral Hemorrhage in Mice.
Translational stroke research
Fu X,Zeng H,Zhao J,Zhou G,Zhou H,Zhuang J,Xu C,Li J,Peng Y,Cao Y,Li Y,Chen H,Wang L,Yan F,Chen G
Wed Dec 01 00:00:00 EST 2021
Inhibition of Dectin-1 Ameliorates Neuroinflammation by Regulating Microglia/Macrophage Phenotype After Intracerebral Hemorrhage in Mice.
Translational stroke research
Fu X,Zeng H,Zhao J,Zhou G,Zhou H,Zhuang J,Xu C,Li J,Peng Y,Cao Y,Li Y,Chen H,Wang L,Yan F,Chen G
Wed Dec 01 00:00:00 EST 2021
Myeloid TBK1 Deficiency Induces Motor Deficits and Axon Degeneration Through Inflammatory Cell Infiltration.
Molecular neurobiology
Duan W,Yi L,Tian Y,Huang HP,Li Z,Bi Y,Guo M,Li Y,Liu Y,Ma Y,Song X,Liu Y,Li C
Sat May 01 00:00:00 EDT 2021
Myeloid TBK1 Deficiency Induces Motor Deficits and Axon Degeneration Through Inflammatory Cell Infiltration.
Molecular neurobiology
Duan W,Yi L,Tian Y,Huang HP,Li Z,Bi Y,Guo M,Li Y,Liu Y,Ma Y,Song X,Liu Y,Li C
Sat May 01 00:00:00 EDT 2021
Probiotic Bacillus cereus Alleviates Dextran Sulfate Sodium-Induced Colitis in Mice through Improvement of the Intestinal Barrier Function, Anti-Inflammation, and Gut Microbiota Modulation.
Journal of agricultural and food chemistry
Sheng K,Xu Y,Kong X,Wang J,Zha X,Wang Y
Wed Dec 15 00:00:00 EST 2021
FNDC5 induces M2 macrophage polarization and promotes hepatocellular carcinoma cell growth by affecting the PPARγ/NF-κB/NLRP3 pathway.
Biochemical and biophysical research communications
Liu H,Wang M,Jin Z,Sun D,Zhu T,Liu X,Tan X,Shi G
Fri Dec 10 00:00:00 EST 2021
Simvastatin Enhances Muscle Regeneration Through Autophagic Defect-Mediated Inflammation and mTOR Activation in G93ASOD1 Mice.
Molecular neurobiology
Wang Y,Bai L,Li S,Wen Y,Liu Q,Li R,Liu Y
Thu Apr 01 00:00:00 EDT 2021
Intracellular Sphingosine-1-Phosphate Receptor 3 Contributes to Lung Tumor Cell Proliferation.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Terlizzi M,Colarusso C,Ferraro G,Monti MC,Rosa I,Somma P,Salvi R,Pinto A,Sorrentino R
Fri Sep 03 00:00:00 EDT 2021
Matrix stiffness regulates bone repair by modulating 12-lipoxygenase-mediated early inflammation.
Materials science & engineering. C, Materials for biological applications
Yao D,Qiao F,Song C,Lv Y
Wed Sep 01 00:00:00 EDT 2021
Matrix stiffness regulates bone repair by modulating 12-lipoxygenase-mediated early inflammation.
Materials science & engineering. C, Materials for biological applications
Yao D,Qiao F,Song C,Lv Y
Wed Sep 01 00:00:00 EDT 2021
Urea as a By-Product of Ammonia Metabolism Can Be a Potential Serum Biomarker of Hepatocellular Carcinoma.
Frontiers in cell and developmental biology
Bai C,Wang H,Dong D,Li T,Yu Z,Guo J,Zhou W,Li D,Yan R,Wang L,Wang Z,Li Y,Ren L
Wed Apr 21 00:00:00 EDT 2021
Suppressing Sart1 to modulate macrophage polarization by siRNA-loaded liposomes: a promising therapeutic strategy for pulmonary fibrosis.
Theranostics
Pan T,Zhou Q,Miao K,Zhang L,Wu G,Yu J,Xu Y,Xiong W,Li Y,Wang Y
Thu Jul 22 00:00:00 EDT 2021
The Protective Role of Yin-Yang 1 in Cardiac Injury and Remodeling After Myocardial Infarction.
Journal of the American Heart Association
Huang Y,Li L,Chen H,Liao Q,Yang X,Yang D,Xia X,Wang H,Wang WE,Chen L,Zeng C
Tue Nov 02 00:00:00 EDT 2021
Pharmacological Activation of RXR-α Promotes Hematoma Absorption via a PPAR-γ-dependent Pathway After Intracerebral Hemorrhage.
Neuroscience bulletin
Xu C,Chen H,Zhou S,Sun C,Xia X,Peng Y,Zhuang J,Fu X,Zeng H,Zhou H,Cao Y,Yu Q,Li Y,Hu L,Zhou G,Yan F,Chen G,Li J
Fri Oct 01 00:00:00 EDT 2021
Pharmacological Activation of RXR-α Promotes Hematoma Absorption via a PPAR-γ-dependent Pathway After Intracerebral Hemorrhage.
Neuroscience bulletin
Xu C,Chen H,Zhou S,Sun C,Xia X,Peng Y,Zhuang J,Fu X,Zeng H,Zhou H,Cao Y,Yu Q,Li Y,Hu L,Zhou G,Yan F,Chen G,Li J
Fri Oct 01 00:00:00 EDT 2021
Amniotic fluid-derived multipotent stromal cells drive diabetic wound healing through modulation of macrophages.
Journal of translational medicine
Subhan BS,Kwong J,Kuhn JF,Monas A,Sharma S,Rabbani PS
Wed Jan 06 00:00:00 EST 2021
Metformin Alleviates Neuroinflammation Following Intracerebral Hemorrhage in Mice by Regulating Microglia/Macrophage Phenotype in a Gut Microbiota-Dependent Manner.
Frontiers in cellular neuroscience
Yu X,Fu X,Wu X,Tang W,Xu L,Hu L,Xu C,Zhou H,Zhou G,Li J,Cao S,Liu J,Yan F,Wang L,Liu F,Chen G
Sat Feb 05 00:00:00 EST 2022
Agomelatine Prevents Amyloid Plaque Deposition, Tau Phosphorylation, and Neuroinflammation in APP/PS1 Mice.
Frontiers in aging neuroscience
Yang XB,Zu HB,Zhao YF,Yao K
Tue Feb 15 00:00:00 EST 2022
Loganin alleviates sepsis-induced acute lung injury by regulating macrophage polarization and inhibiting NLRP3 inflammasome activation.
International immunopharmacology
Zhang J,Wang C,Wang H,Li X,Xu J,Yu K
Tue Jun 01 00:00:00 EDT 2021
Loss of Diacylglycerol Kinase α Enhances Macrophage Responsiveness.
Frontiers in immunology
Manigat LC,Granade ME,Taori S,Miller CA,Vass LR,Zhong XP,Harris TE,Purow BW
Fri Jan 07 00:00:00 EST 2022
PAK1 Silencing Attenuated Proinflammatory Macrophage Activation and Foam Cell Formation by Increasing PPARγ Expression.
Oxidative medicine and cellular longevity
Cheng WL,Zhang Q,Li B,Cao JL,Jiao L,Chao SP,Lu Z,Zhao F
Mon Jan 31 00:00:00 EST 2022
The Impact of β-1,4-Galactosyltransferase V on Microglial Function.
Frontiers in cellular neuroscience
Wang X,Shi N,Hui M,Jin H,Gao S,Zhou Q,Zhang L,Yan M,Shen H
Tue Sep 21 00:00:00 EDT 2021
Distinct phenotypic expression levels of macrophages in neonatal lungs.
Experimental and therapeutic medicine
Fang SY,Chen JL,Chiu MH,Huang CC,Lin MW,Lam CF
Thu Apr 01 00:00:00 EDT 2021
Preventive and Therapeutic Spermidine Treatment Attenuates Acute Colitis in Mice.
Journal of agricultural and food chemistry
Ma L,Ni L,Yang T,Mao P,Huang X,Luo Y,Jiang Z,Hu L,Zhao Y,Fu Z,Ni Y
Wed Feb 17 00:00:00 EST 2021
Saponins from Panax japonicus attenuate cognitive impairment in ageing rats through regulating microglial polarisation and autophagy.
Pharmaceutical biology
Pi XJ,Zhao QQ,Wang JX,Zhang XL,Yuan D,Hu SS,He YM,Zhang CC,Zhou ZY,Wang T
Wed Dec 01 00:00:00 EST 2021
PD-L1 Improves Motor Function and Alleviates Neuropathic Pain in Male Mice After Spinal Cord Injury by Inhibiting MAPK Pathway.
Frontiers in immunology
Kong F,Sun K,Zhu J,Li F,Lin F,Sun X,Luo X,Ren C,Lu L,Zhao S,Sun J,Wang Y,Shi J
Mon Nov 01 00:00:00 EDT 2021
PD-L1 Improves Motor Function and Alleviates Neuropathic Pain in Male Mice After Spinal Cord Injury by Inhibiting MAPK Pathway.
Frontiers in immunology
Kong F,Sun K,Zhu J,Li F,Lin F,Sun X,Luo X,Ren C,Lu L,Zhao S,Sun J,Wang Y,Shi J
Mon Nov 01 00:00:00 EDT 2021
MCC950 Ameliorates Acute Liver Injury Through Modulating Macrophage Polarization and Myeloid-Derived Suppressor Cells Function.
Frontiers in medicine
Yan W,Shen Y,Huang J,Lu L,Zhang Q
Tue Dec 07 00:00:00 EST 2021
A Novel Hydrogen Sulfide Donor Reduces Pilocarpine-Induced Status Epilepticus and Regulates Microglial Inflammatory Profile.
Frontiers in cellular neuroscience
Liu Z,Zhu Z,He Y,Kang Q,Li F,Zhang W,He Y,Lin Y,Huang B,Mo M,Xu P,Zhu X
Tue Dec 21 00:00:00 EST 2021
A Novel Hydrogen Sulfide Donor Reduces Pilocarpine-Induced Status Epilepticus and Regulates Microglial Inflammatory Profile.
Frontiers in cellular neuroscience
Liu Z,Zhu Z,He Y,Kang Q,Li F,Zhang W,He Y,Lin Y,Huang B,Mo M,Xu P,Zhu X
Tue Dec 21 00:00:00 EST 2021
Inhibition of P2X4R attenuates white matter injury in mice after intracerebral hemorrhage by regulating microglial phenotypes.
Journal of neuroinflammation
Fu X,Zhou G,Wu X,Xu C,Zhou H,Zhuang J,Peng Y,Cao Y,Zeng H,Li Y,Li J,Gao L,Chen G,Wang L,Yan F
Mon Aug 23 00:00:00 EDT 2021
LncRNA LINC01088 inhibits the function of trophoblast cells, activates the MAPK-signaling pathway and associates with recurrent pregnancy loss.
Molecular human reproduction
Zhao H,Li Y,Dong N,Zhang L,Chen X,Mao H,Al-Ameri SAA,Wang X,Wang Q,Du L,Wang C,Mao H
Sat Aug 07 00:00:00 EDT 2021
Effective-compound combination inhibits the M2-like polarization of macrophages and attenuates the development of pulmonary fibrosis by increasing autophagy through mTOR signaling.
International immunopharmacology
Zhao P,Cai Z,Tian Y,Li J,Li K,Li M,Bai Y,Li J
Wed Dec 01 00:00:00 EST 2021
Effective-compound combination inhibits the M2-like polarization of macrophages and attenuates the development of pulmonary fibrosis by increasing autophagy through mTOR signaling.
International immunopharmacology
Zhao P,Cai Z,Tian Y,Li J,Li K,Li M,Bai Y,Li J
Wed Dec 01 00:00:00 EST 2021
Colon-Targeted Adhesive Hydrogel Microsphere for Regulation of Gut Immunity and Flora.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu H,Cai Z,Wang F,Hong L,Deng L,Zhong J,Wang Z,Cui W
Wed Sep 01 00:00:00 EDT 2021
Colon-Targeted Adhesive Hydrogel Microsphere for Regulation of Gut Immunity and Flora.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu H,Cai Z,Wang F,Hong L,Deng L,Zhong J,Wang Z,Cui W
Wed Sep 01 00:00:00 EDT 2021
Schizandrin A ameliorates cognitive functions via modulating microglial polarisation in Alzheimer's disease mice.
Pharmaceutical biology
Wang Q,Liu L,Guan H,Zhou Y,Li Q
Wed Dec 01 00:00:00 EST 2021
lncRNA Gm16410 Mediates PM2.5-Induced Macrophage Activation via PI3K/AKT Pathway.
Frontiers in cell and developmental biology
Xu J,Xu H,Ma K,Wang Y,Niu B,Zhang L,Li F
Sat Apr 03 00:00:00 EDT 2021
M2 Macrophage-Derived Exosomal miR-590-3p Attenuates DSS-Induced Mucosal Damage and Promotes Epithelial Repair via the LATS1/YAP/ β-Catenin Signalling Axis.
Journal of Crohn's & colitis
Deng F,Yan J,Lu J,Luo M,Xia P,Liu S,Wang X,Zhi F,Liu D
Tue Apr 06 00:00:00 EDT 2021
Asiatic acid attenuates diabetic retinopathy through TLR4/MyD88/NF-κB p65 mediated modulation of microglia polarization.
Life sciences
Fang M,Wan W,Li Q,Wan W,Long Y,Liu H,Yang X
Thu Jul 15 00:00:00 EDT 2021
Regulatory T cells protect against brain damage by alleviating inflammatory response in neuromyelitis optica spectrum disorder.
Journal of neuroinflammation
Ma X,Qin C,Chen M,Yu HH,Chu YH,Chen TJ,Bosco DB,Wu LJ,Bu BT,Wang W,Tian DS
Wed Sep 15 00:00:00 EDT 2021
Regulatory T cells protect against brain damage by alleviating inflammatory response in neuromyelitis optica spectrum disorder.
Journal of neuroinflammation
Ma X,Qin C,Chen M,Yu HH,Chu YH,Chen TJ,Bosco DB,Wu LJ,Bu BT,Wang W,Tian DS
Wed Sep 15 00:00:00 EDT 2021
Interleukin-19 Abrogates Experimental Autoimmune Encephalomyelitis by Attenuating Antigen-Presenting Cell Activation.
Frontiers in immunology
Horiuchi H,Parajuli B,Komiya H,Ogawa Y,Jin S,Takahashi K,Azuma YT,Tanaka F,Suzumura A,Takeuchi H
Tue Jun 29 00:00:00 EDT 2021
Vagus nerve stimulation mediates microglia M1/2 polarization via inhibition of TLR4 pathway after ischemic stroke.
Biochemical and biophysical research communications
Zhang L,Liu Y,Wang S,Long L,Zang Q,Ma J,Yu L,Jia G
Fri Nov 05 00:00:00 EDT 2021
Reprogramming immunosuppressive myeloid cells facilitates immunotherapy for colorectal cancer.
EMBO molecular medicine
Lu W,Yu W,He J,Liu W,Yang J,Lin X,Zhang Y,Wang X,Jiang W,Luo J,Zhang Q,Yang H,Peng S,Yi Z,Ren S,Chen J,Siwko S,Nussinov R,Cheng F,Zhang H,Liu M
Mon Jan 11 00:00:00 EST 2021
Reprogramming immunosuppressive myeloid cells facilitates immunotherapy for colorectal cancer.
EMBO molecular medicine
Lu W,Yu W,He J,Liu W,Yang J,Lin X,Zhang Y,Wang X,Jiang W,Luo J,Zhang Q,Yang H,Peng S,Yi Z,Ren S,Chen J,Siwko S,Nussinov R,Cheng F,Zhang H,Liu M
Mon Jan 11 00:00:00 EST 2021
Ameliorative effects of miR-423-5p against polarization of microglial cells of the M1 phenotype by targeting a NLRP3 inflammasome signaling pathway.
International immunopharmacology
Cheng J,Hao J,Jiang X,Ji J,Wu T,Chen X,Zhang F
Fri Oct 01 00:00:00 EDT 2021
Ameliorative effects of miR-423-5p against polarization of microglial cells of the M1 phenotype by targeting a NLRP3 inflammasome signaling pathway.
International immunopharmacology
Cheng J,Hao J,Jiang X,Ji J,Wu T,Chen X,Zhang F
Fri Oct 01 00:00:00 EDT 2021
Myeloid NEMO deficiency promotes tumor immunosuppression partly via MCP1-CCR2 axis.
Experimental cell research
Yuanyuan L,Yakun L,Zhongyao L,Le Y,Weisong D,Moran G,Hui B,Chunyan L
Mon Feb 15 00:00:00 EST 2021
β2-Adrenergic Receptor Enhances the Alternatively Activated Macrophages and Promotes Biliary Injuries Caused by Helminth Infection.
Frontiers in immunology
Koda S,Zhang B,Zhou QY,Xu N,Li J,Liu JX,Liu M,Lv ZY,Wang JL,Shi Y,Gao S,Yu Q,Li XY,Xu YH,Chen JX,Tekengne BOT,Adzika GK,Tang RX,Sun H,Zheng KY,Yan C
Fri Feb 11 00:00:00 EST 2022
Aminooxyacetic acid attenuates post-infarct cardiac dysfunction by balancing macrophage polarization through modulating macrophage metabolism in mice.
Journal of cellular and molecular medicine
Zhao P,Zhou W,Zhang Y,Li J,Zhao Y,Pan L,Shen Z,Chen W,Hui J
Sat Feb 01 00:00:00 EST 2020
Aminooxyacetic acid attenuates post-infarct cardiac dysfunction by balancing macrophage polarization through modulating macrophage metabolism in mice.
Journal of cellular and molecular medicine
Zhao P,Zhou W,Zhang Y,Li J,Zhao Y,Pan L,Shen Z,Chen W,Hui J
Sat Feb 01 00:00:00 EST 2020
Blocking P2X7-Mediated Macrophage Polarization Overcomes Treatment Resistance in Lung Cancer.
Cancer immunology research
Qin J,Zhang X,Tan B,Zhang S,Yin C,Xue Q,Zhang Z,Ren H,Chen J,Liu M,Qian M,Du B
Sun Nov 01 00:00:00 EDT 2020
An extracorporeal bioartificial liver embedded with 3D-layered human liver progenitor-like cells relieves acute liver failure in pigs.
Science translational medicine
Li WJ,Zhu XJ,Yuan TJ,Wang ZY,Bian ZQ,Jing HS,Shi X,Chen CY,Fu GB,Huang WJ,Shi YP,Liu Q,Zeng M,Zhang HD,Wu HP,Yu WF,Zhai B,Yan HX
Wed Jul 08 00:00:00 EDT 2020
Advanced glycation end products enhance macrophage polarization to the M1 phenotype via the HIF-1α/PDK4 pathway.
Molecular and cellular endocrinology
Han X,Ma W,Zhu Y,Sun X,Liu N
Thu Aug 20 00:00:00 EDT 2020
Advanced glycation end products enhance macrophage polarization to the M1 phenotype via the HIF-1α/PDK4 pathway.
Molecular and cellular endocrinology
Han X,Ma W,Zhu Y,Sun X,Liu N
Thu Aug 20 00:00:00 EDT 2020
Fucoidan inhibits tooth movement by promoting restorative macrophage polarization through the STAT3 pathway.
Journal of cellular physiology
Zhang S,Zhang H,Jin Z,Wang S,Wang Y,Zhu L,Sun W,Yan B
Tue Sep 01 00:00:00 EDT 2020
Csf2 Attenuated Sepsis-Induced Acute Kidney Injury by Promoting Alternative Macrophage Transition.
Frontiers in immunology
Li Y,Zhai P,Zheng Y,Zhang J,Kellum JA,Peng Z
Tue Apr 06 00:00:00 EDT 2021
Rosmarinic acid exerts a neuroprotective effect on spinal cord injury by suppressing oxidative stress and inflammation via modulating the Nrf2/HO-1 and TLR4/NF-κB pathways.
Toxicology and applied pharmacology
Ma Z,Lu Y,Yang F,Li S,He X,Gao Y,Zhang G,Ren E,Wang Y,Kang X
Sun Apr 19 00:00:00 EDT 2020
An injectable recombinant human milk fat globule-epidermal growth factor 8-loaded copolymer system for spinal cord injury reduces inflammation through NF-κB and neuronal cell death.
Cytotherapy
Gong Z,Wang C,Ni L,Ying L,Shu J,Wang J,Yu C,Xia K,Cheng F,Shi K,Xu G,Yu Q,Shen J,Chen Q,Li F,Liang C
Wed Apr 01 00:00:00 EDT 2020
Inhibition of miR-181a-5p reduces astrocyte and microglia activation and oxidative stress by activating SIRT1 in immature rats with epilepsy.
Laboratory investigation; a journal of technical methods and pathology
Kong H,Wang H,Zhuo Z,Li Z,Tian P,Wu J,Liu J,Chen Z,Zhang J,Luo Q
Tue Sep 01 00:00:00 EDT 2020
AhR activation attenuates calcium oxalate nephrocalcinosis by diminishing M1 macrophage polarization and promoting M2 macrophage polarization.
Theranostics
Yang X,Liu H,Ye T,Duan C,Lv P,Wu X,Liu J,Jiang K,Lu H,Yang H,Xia D,Peng E,Chen Z,Tang K,Ye Z
Tue Jun 01 00:00:00 EDT 2021
Ketogenic Diet Elicits Antitumor Properties through Inducing Oxidative Stress, Inhibiting MMP-9 Expression, and Rebalancing M1/M2 Tumor-Associated Macrophage Phenotype in a Mouse Model of Colon Cancer.
Journal of agricultural and food chemistry
Zhang N,Liu C,Jin L,Zhang R,Wang T,Wang Q,Chen J,Yang F,Siebert HC,Zheng X
Wed Oct 07 00:00:00 EDT 2020
TNF-α/HMGB1 inflammation signalling pathway regulates pyroptosis during liver failure and acute kidney injury.
Cell proliferation
Wang Y,Zhang H,Chen Q,Jiao F,Shi C,Pei M,Lv J,Zhang H,Wang L,Gong Z
Mon Jun 01 00:00:00 EDT 2020
The effect of combining Endostar with radiotherapy on blood vessels, tumor-associated macrophages, and T cells in brain metastases of Lewis lung cancer.
Translational lung cancer research
Peng L,Wang Y,Fei S,Wei C,Tong F,Wu G,Ma H,Dong X
Mon Jun 01 00:00:00 EDT 2020
Hypothermia-Induced Ubiquitination of Voltage-Dependent Anion Channel 3 Protects BV2 Microglia Cells From Cytotoxicity Following Oxygen-Glucose Deprivation/Recovery.
Frontiers in molecular neuroscience
Zhao S,Xiao P,Cui H,Gong P,Lin C,Chen F,Tang Z
Mon Sep 28 00:00:00 EDT 2020
Relationship of tumour-associated macrophages with poor prognosis in Wilms' tumour.
Journal of pediatric urology
Tian K,Wang X,Wu Y,Wu X,Du G,Liu W,Wu R
Mon Jun 01 00:00:00 EDT 2020
Disease Stage-Associated Alterations in Learning and Memory through the Electroacupuncture Modulation of the Cortical Microglial M1/M2 Polarization in Mice with Alzheimer's Disease.
Neural plasticity
Li L,Li L,Zhang J,Huang S,Liu W,Wang Z,Liang S,Tao J,Chen L
Tue Aug 03 00:00:00 EDT 2021
Analysis of Cyclooxygenase 2, Programmed Cell Death Ligand 1, and Arginase 1 Expression in Human Pituitary Adenoma.
World neurosurgery
Zhao G,Chen W,He J,Cui C,Zhao L,Zhao Y,Sun C,Nie D,Jin F,Kong L
Tue Dec 01 00:00:00 EST 2020
MG53 Protects hUC-MSCs against Inflammatory Damage and Synergistically Enhances Their Efficacy in Neuroinflammation Injured Brain through Inhibiting NLRP3/Caspase-1/IL-1β Axis.
ACS chemical neuroscience
Ma S,Wang Y,Zhou X,Li Z,Zhang Z,Wang Y,Huang T,Zhang Y,Shi J,Guan F
Wed Sep 02 00:00:00 EDT 2020
SWI/SNF complex subunit BAF60a represses hepatic ureagenesis through a crosstalk between YB-1 and PGC-1α.
Molecular metabolism
Zhang W,Dong Z,Xu M,Zhang S,Liu C,Chen S
Sat Feb 01 00:00:00 EST 2020
Therapeutic Role of Protein Tyrosine Phosphatase 1B in Parkinson's Disease via Antineuroinflammation and Neuroprotection In Vitro and In Vivo.
Parkinson's disease
Feng CW,Chen NF,Chan TF,Chen WF
Tue Jan 19 00:00:00 EST 2021
Functional changes of the liver in the absence of growth hormone (GH) action - Proteomic and metabolomic insights from a GH receptor deficient pig model.
Molecular metabolism
Riedel EO,Hinrichs A,Kemter E,Dahlhoff M,Backman M,Rathkolb B,Prehn C,Adamski J,Renner S,Blutke A,de Angelis MH,Bidlingmaier M,Schopohl J,Arnold GJ,Fröhlich T,Wolf E
Mon Jun 01 00:00:00 EDT 2020
Beneficial Effects of Theta-Burst Transcranial Magnetic Stimulation on Stroke Injury via Improving Neuronal Microenvironment and Mitochondrial Integrity.
Translational stroke research
Zong X,Dong Y,Li Y,Yang L,Li Y,Yang B,Tucker L,Zhao N,Brann DW,Yan X,Hu S,Zhang Q
Mon Jun 01 00:00:00 EDT 2020
Anti-arthritis effect of berberine associated with regulating energy metabolism of macrophages through AMPK/ HIF-1α pathway.
International immunopharmacology
Yu Y,Cai W,Zhou J,Lu H,Wang Y,Song Y,He R,Pei F,Wang X,Zhang R,Liu H,Wei F
Thu Oct 01 00:00:00 EDT 2020
Anti-arthritis effect of berberine associated with regulating energy metabolism of macrophages through AMPK/ HIF-1α pathway.
International immunopharmacology
Yu Y,Cai W,Zhou J,Lu H,Wang Y,Song Y,He R,Pei F,Wang X,Zhang R,Liu H,Wei F
Thu Oct 01 00:00:00 EDT 2020
Alternatively activated macrophages are associated with the α2AP production that occurs with the development of dermal fibrosis : The role of alternatively activated macrophages on the development of fibrosis.
Arthritis research & therapy
Kanno Y,Shu E,Niwa H,Kanoh H,Seishima M
Fri Apr 10 00:00:00 EDT 2020
Sulforaphane elicts dual therapeutic effects on Renal Inflammatory Injury and crystal deposition in Calcium Oxalate Nephrocalcinosis.
Theranostics
Liu H,Yang X,Tang K,Ye T,Duan C,Lv P,Yan L,Wu X,Chen Z,Liu J,Deng Y,Zeng G,Xing J,Ye Z,Xu H
Mon May 03 00:00:00 EDT 2021
Keratinocyte-Macrophage Crosstalk by the Nrf2/Ccl2/EGF Signaling Axis Orchestrates Tissue Repair.
Cell reports
Villarreal-Ponce A,Tiruneh MW,Lee J,Guerrero-Juarez CF,Kuhn J,David JA,Dammeyer K,Mc Kell R,Kwong J,Rabbani PS,Nie Q,Ceradini DJ
Tue Nov 24 00:00:00 EST 2020
Herb-sourced emodin inhibits angiogenesis of breast cancer by targeting VEGFA transcription.
Theranostics
Zou G,Zhang X,Wang L,Li X,Xie T,Zhao J,Yan J,Wang L,Ye H,Jiao S,Xiang R,Shi Y
Tue May 18 00:00:00 EDT 2021
Bone marrow mesenchymal stem cell-derived exosomes prevent osteoarthritis by regulating synovial macrophage polarization.
Aging
Zhang J,Rong Y,Luo C,Cui W
Tue Dec 22 00:00:00 EST 2020
Bone marrow mesenchymal stem cell-derived exosomes prevent osteoarthritis by regulating synovial macrophage polarization.
Aging
Zhang J,Rong Y,Luo C,Cui W
Tue Dec 22 00:00:00 EST 2020
Bone marrow mesenchymal stem cell-derived exosomes prevent osteoarthritis by regulating synovial macrophage polarization.
Aging
Zhang J,Rong Y,Luo C,Cui W
Tue Dec 22 00:00:00 EST 2020
Bone marrow mesenchymal stem cell-derived exosomes prevent osteoarthritis by regulating synovial macrophage polarization.
Aging
Zhang J,Rong Y,Luo C,Cui W
Tue Dec 22 00:00:00 EST 2020
A free-standing multilayer film as a novel delivery carrier of platelet lysates for potential wound-dressing applications.
Biomaterials
Tang Q,Lim T,Wei XJ,Wang QY,Xu JC,Shen LY,Zhu ZZ,Zhang CQ
Thu Oct 01 00:00:00 EDT 2020
Fusobacterium nucleatum Aggravates the Progression of Colitis by Regulating M1 Macrophage Polarization via AKT2 Pathway.
Frontiers in immunology
Liu L,Liang L,Liang H,Wang M,Lu B,Xue M,Deng J,Chen Y
Tue Oct 20 00:00:00 EDT 2020
Fusobacterium nucleatum Aggravates the Progression of Colitis by Regulating M1 Macrophage Polarization via AKT2 Pathway.
Frontiers in immunology
Liu L,Liang L,Liang H,Wang M,Lu B,Xue M,Deng J,Chen Y
Tue Oct 20 00:00:00 EDT 2020
Targeted Delivery of miRNA 155 to Tumor Associated Macrophages for Tumor Immunotherapy.
Molecular pharmaceutics
Zang X,Zhang X,Zhao X,Hu H,Qiao M,Deng Y,Chen D
Mon Apr 01 00:00:00 EDT 2019
Pancreatic cancer-educated macrophages protect cancer cells from complement-dependent cytotoxicity by up-regulation of CD59.
Cell death & disease
Zhang R,Liu Q,Peng J,Wang M,Gao X,Liao Q,Zhao Y
Mon Nov 04 00:00:00 EST 2019
Comparative Proteomic Analysis of Murine Cutaneous Lesions Induced by Leishmania amazonensis or Leishmania major.
ACS infectious diseases
Negrão F,Giorgio S,Eberlin MN,Yates JR
Fri Aug 09 00:00:00 EDT 2019
Static Magnetic Field Accelerates Diabetic Wound Healing by Facilitating Resolution of Inflammation.
Journal of diabetes research
Shang W,Chen G,Li Y,Zhuo Y,Wang Y,Fang Z,Yu Y,Ren H
Tue Jun 09 00:00:00 EDT 2020
Role of Tim-3 in Decidual Macrophage Functional Polarization During Abnormal Pregnancy With Toxoplasma gondii Infection.
Frontiers in immunology
Zhang D,Ren L,Zhao M,Yang C,Liu X,Zhang H,Jiang Y,Sun X,Li T,Cui L,Hu X
Thu Oct 15 00:00:00 EDT 2020
Berberine attenuates arthritis in adjuvant-induced arthritic rats associated with regulating polarization of macrophages through AMPK/NF-кB pathway.
European journal of pharmacology
Zhou J,Yu Y,Yang X,Wang Y,Song Y,Wang Q,Chen Z,Zong S,Fan M,Meng X,Xie C,Zhou F,Liu H,Wei F
Wed Jun 05 00:00:00 EDT 2019
Berberine attenuates arthritis in adjuvant-induced arthritic rats associated with regulating polarization of macrophages through AMPK/NF-кB pathway.
European journal of pharmacology
Zhou J,Yu Y,Yang X,Wang Y,Song Y,Wang Q,Chen Z,Zong S,Fan M,Meng X,Xie C,Zhou F,Liu H,Wei F
Wed Jun 05 00:00:00 EDT 2019
Myeloid-Derived Suppressor Cells and γδT17 Cells Contribute to the Development of Gastric MALT Lymphoma in H. felis-Infected Mice.
Frontiers in immunology
Zhao Y,Lu F,Ye J,Ji M,Pang Y,Wang Y,Wang L,Li G,Sun T,Li J,Ma D,Ji C
Mon Nov 16 00:00:00 EST 2020
Significant Down-Regulation of Urea Cycle Generates Clinically Relevant Proteomic Signature in Hepatocellular Carcinoma Patients with Macrovascular Invasion.
Journal of proteome research
Cao Y,Ding W,Zhang J,Gao Q,Yang H,Cao W,Wang Z,Fang L,Du R
Fri May 03 00:00:00 EDT 2019
Interleukin-1 receptor associated kinase (IRAK)-M -mediated type 2 microglia polarization ameliorates the severity of experimental autoimmune encephalomyelitis (EAE).
Journal of autoimmunity
Liu B,Gu Y,Pei S,Peng Y,Chen J,Pham LV,Shen HY,Zhang J,Wang H
Thu Aug 01 00:00:00 EDT 2019
Interleukin-1 receptor associated kinase (IRAK)-M -mediated type 2 microglia polarization ameliorates the severity of experimental autoimmune encephalomyelitis (EAE).
Journal of autoimmunity
Liu B,Gu Y,Pei S,Peng Y,Chen J,Pham LV,Shen HY,Zhang J,Wang H
Thu Aug 01 00:00:00 EDT 2019
Toxoplasma ROP16I/III ameliorated inflammatory bowel diseases via inducing M2 phenotype of macrophages.
World journal of gastroenterology
Xu YW,Xing RX,Zhang WH,Li L,Wu Y,Hu J,Wang C,Luo QL,Shen JL,Chen X
Sat Dec 07 00:00:00 EST 2019
The SIAH2-NRF1 axis spatially regulates tumor microenvironment remodeling for tumor progression.
Nature communications
Ma B,Cheng H,Mu C,Geng G,Zhao T,Luo Q,Ma K,Chang R,Liu Q,Gao R,Nie J,Xie J,Han J,Chen L,Ma G,Zhu Y,Chen Q
Mon Mar 04 00:00:00 EST 2019
Tandem Mass Tag Proteomic Analysis of in Vitro and in Vivo Models of Cutaneous Leishmaniasis Reveals Parasite-Specific and Nonspecific Modulation of Proteins in the Host.
ACS infectious diseases
Negrão F,Diedrich JK,Giorgio S,Eberlin MN,Yates JR
Fri Dec 13 00:00:00 EST 2019
Metformin enhances gefitinib efficacy by interfering with interactions between tumor-associated macrophages and head and neck squamous cell carcinoma cells.
Cellular oncology (Dordrecht)
Yin X,Han S,Song C,Zou H,Wei Z,Xu W,Ran J,Tang C,Wang Y,Cai Y,Hu Q,Han W
Thu Aug 01 00:00:00 EDT 2019
Metformin enhances gefitinib efficacy by interfering with interactions between tumor-associated macrophages and head and neck squamous cell carcinoma cells.
Cellular oncology (Dordrecht)
Yin X,Han S,Song C,Zou H,Wei Z,Xu W,Ran J,Tang C,Wang Y,Cai Y,Hu Q,Han W
Thu Aug 01 00:00:00 EDT 2019
Macrophage polarization in periodontal ligament stem cells enhanced periodontal regeneration.
Stem cell research & therapy
Liu J,Chen B,Bao J,Zhang Y,Lei L,Yan F
Fri Nov 15 00:00:00 EST 2019
PTPN2 improved renal injury and fibrosis by suppressing STAT-induced inflammation in early diabetic nephropathy.
Journal of cellular and molecular medicine
Li Y,Zhou H,Li Y,Han L,Song M,Chen F,Shang G,Wang D,Wang Z,Zhang W,Zhong M
Sat Jun 01 00:00:00 EDT 2019
PTPN2 improved renal injury and fibrosis by suppressing STAT-induced inflammation in early diabetic nephropathy.
Journal of cellular and molecular medicine
Li Y,Zhou H,Li Y,Han L,Song M,Chen F,Shang G,Wang D,Wang Z,Zhang W,Zhong M
Sat Jun 01 00:00:00 EDT 2019
Aryl Hydrocarbon Receptor Regulates Apoptosis and Inflammation in a Murine Model of Experimental Autoimmune Uveitis.
Frontiers in immunology
Huang Y,He J,Liang H,Hu K,Jiang S,Yang L,Mei S,Zhu X,Yu J,Kijlstra A,Yang P,Hou S
Wed Nov 20 00:00:00 EST 2019
Aryl Hydrocarbon Receptor Regulates Apoptosis and Inflammation in a Murine Model of Experimental Autoimmune Uveitis.
Frontiers in immunology
Huang Y,He J,Liang H,Hu K,Jiang S,Yang L,Mei S,Zhu X,Yu J,Kijlstra A,Yang P,Hou S
Wed Nov 20 00:00:00 EST 2019
Effective Amelioration of Liver Fibrosis Through Lentiviral Vector Carrying in Murine Model.
Frontiers in immunology
Liu L,Jin M,Tao Q,Yu L,Du J,Wang C,Luo Q,Xing T,Xu Y,Shen J,Chu D
Wed Nov 20 00:00:00 EST 2019
Effective Amelioration of Liver Fibrosis Through Lentiviral Vector Carrying in Murine Model.
Frontiers in immunology
Liu L,Jin M,Tao Q,Yu L,Du J,Wang C,Luo Q,Xing T,Xu Y,Shen J,Chu D
Wed Nov 20 00:00:00 EST 2019
Photobiomodulation therapy promotes neurogenesis by improving post-stroke local microenvironment and stimulating neuroprogenitor cells.
Experimental neurology
Yang L,Tucker D,Dong Y,Wu C,Lu Y,Li Y,Zhang J,Liu TC,Zhang Q
Mon Jan 01 00:00:00 EST 2018
Photobiomodulation therapy promotes neurogenesis by improving post-stroke local microenvironment and stimulating neuroprogenitor cells.
Experimental neurology
Yang L,Tucker D,Dong Y,Wu C,Lu Y,Li Y,Zhang J,Liu TC,Zhang Q
Mon Jan 01 00:00:00 EST 2018
l-Arginine induces antioxidant response to prevent oxidative stress via stimulation of glutathione synthesis and activation of Nrf2 pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Liang M,Wang Z,Li H,Cai L,Pan J,He H,Wu Q,Tang Y,Ma J,Yang L
Tue May 01 00:00:00 EDT 2018
l-Arginine induces antioxidant response to prevent oxidative stress via stimulation of glutathione synthesis and activation of Nrf2 pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Liang M,Wang Z,Li H,Cai L,Pan J,He H,Wu Q,Tang Y,Ma J,Yang L
Tue May 01 00:00:00 EDT 2018
ER-localized protein-Herpud1 is a new mediator of IL-4-induced macrophage polarization and migration.
Experimental cell research
Li Y,Xie Y,Hao J,Liu J,Ning Y,Tang Q,Ma M,Zhou H,Guan S,Zhou Q,Lv X
Sun Jul 15 00:00:00 EDT 2018
ER-localized protein-Herpud1 is a new mediator of IL-4-induced macrophage polarization and migration.
Experimental cell research
Li Y,Xie Y,Hao J,Liu J,Ning Y,Tang Q,Ma M,Zhou H,Guan S,Zhou Q,Lv X
Sun Jul 15 00:00:00 EDT 2018
S100A8/A9 induces microglia activation and promotes the apoptosis of oligodendrocyte precursor cells by activating the NF-κB signaling pathway.
Brain research bulletin
Wu M,Xu L,Wang Y,Zhou N,Zhen F,Zhang Y,Qu X,Fan H,Liu S,Chen Y,Yao R
Mon Oct 01 00:00:00 EDT 2018
S100A8/A9 induces microglia activation and promotes the apoptosis of oligodendrocyte precursor cells by activating the NF-κB signaling pathway.
Brain research bulletin
Wu M,Xu L,Wang Y,Zhou N,Zhen F,Zhang Y,Qu X,Fan H,Liu S,Chen Y,Yao R
Mon Oct 01 00:00:00 EDT 2018
Interleukin-33 Protects Ischemic Brain Injury by Regulating Specific Microglial Activities.
Neuroscience
Luo Q,Fan Y,Lin L,Wei J,Li Z,Li Y,Nakae S,Lin W,Chen Q
Fri Aug 10 00:00:00 EDT 2018
Interleukin-33 Protects Ischemic Brain Injury by Regulating Specific Microglial Activities.
Neuroscience
Luo Q,Fan Y,Lin L,Wei J,Li Z,Li Y,Nakae S,Lin W,Chen Q
Fri Aug 10 00:00:00 EDT 2018
Rapamycin and FTY720 Alleviate Atherosclerosis by Cross Talk of Macrophage Polarization and Autophagy.
BioMed research international
Sun RZ,Fan Y,Liang X,Gong TT,Wang Q,Liu H,Shan ZY,Lei L
Wed Apr 24 00:00:00 EDT 2019
Rapamycin and FTY720 Alleviate Atherosclerosis by Cross Talk of Macrophage Polarization and Autophagy.
BioMed research international
Sun RZ,Fan Y,Liang X,Gong TT,Wang Q,Liu H,Shan ZY,Lei L
Wed Apr 24 00:00:00 EDT 2019
Clemastine Alleviates Depressive-Like Behavior Through Reversing the Imbalance of Microglia-Related Pro-inflammatory State in Mouse Hippocampus.
Frontiers in cellular neuroscience
Su WJ,Zhang T,Jiang CL,Wang W
Wed Sep 30 00:00:00 EDT 2020
Clemastine Alleviates Depressive-Like Behavior Through Reversing the Imbalance of Microglia-Related Pro-inflammatory State in Mouse Hippocampus.
Frontiers in cellular neuroscience
Su WJ,Zhang T,Jiang CL,Wang W
Wed Sep 30 00:00:00 EDT 2020
The Role of Alpha-Lipoic Acid in the Pathomechanism of Acute Ischemic Stroke.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Q,Lv C,Sun Y,Han X,Wang S,Mao Z,Xin Y,Zhang B
Tue Sep 04 00:00:00 EDT 2018
The Role of Alpha-Lipoic Acid in the Pathomechanism of Acute Ischemic Stroke.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Q,Lv C,Sun Y,Han X,Wang S,Mao Z,Xin Y,Zhang B
Tue Sep 04 00:00:00 EDT 2018
Targeting MAPK Pathways by Naringenin Modulates Microglia M1/M2 Polarization in Lipopolysaccharide-Stimulated Cultures.
Frontiers in cellular neuroscience
Zhang B,Wei YZ,Wang GQ,Li DD,Shi JS,Zhang F
Thu Oct 01 00:00:00 EDT 2020
Reduced bioavailable manganese causes striatal urea cycle pathology in Huntington's disease mouse model.
Biochimica et biophysica acta. Molecular basis of disease
Bichell TJV,Wegrzynowicz M,Tipps KG,Bradley EM,Uhouse MA,Bryan M,Horning K,Fisher N,Dudek K,Halbesma T,Umashanker P,Stubbs AD,Holt HK,Kwakye GF,Tidball AM,Colbran RJ,Aschner M,Neely MD,Di Pardo A,Maglione V,Osmand A,Bowman AB
Thu Jun 01 00:00:00 EDT 2017
Reduced bioavailable manganese causes striatal urea cycle pathology in Huntington's disease mouse model.
Biochimica et biophysica acta. Molecular basis of disease
Bichell TJV,Wegrzynowicz M,Tipps KG,Bradley EM,Uhouse MA,Bryan M,Horning K,Fisher N,Dudek K,Halbesma T,Umashanker P,Stubbs AD,Holt HK,Kwakye GF,Tidball AM,Colbran RJ,Aschner M,Neely MD,Di Pardo A,Maglione V,Osmand A,Bowman AB
Thu Jun 01 00:00:00 EDT 2017
GRP78 plays an integral role in tumor cell inflammation-related migration induced by M2 macrophages.
Cellular signalling
Zhang L,Li Z,Ding G,La X,Yang P,Li Z
Fri Sep 01 00:00:00 EDT 2017
GRP78 plays an integral role in tumor cell inflammation-related migration induced by M2 macrophages.
Cellular signalling
Zhang L,Li Z,Ding G,La X,Yang P,Li Z
Fri Sep 01 00:00:00 EDT 2017
Treadmill Exercise Exerts Neuroprotection and Regulates Microglial Polarization and Oxidative Stress in a Streptozotocin-Induced Rat Model of Sporadic Alzheimer's Disease.
Journal of Alzheimer's disease : JAD
Lu Y,Dong Y,Tucker D,Wang R,Ahmed ME,Brann D,Zhang Q
Fri Feb 23 00:00:00 EST 2018
Treadmill Exercise Exerts Neuroprotection and Regulates Microglial Polarization and Oxidative Stress in a Streptozotocin-Induced Rat Model of Sporadic Alzheimer's Disease.
Journal of Alzheimer's disease : JAD
Lu Y,Dong Y,Tucker D,Wang R,Ahmed ME,Brann D,Zhang Q
Fri Feb 23 00:00:00 EST 2018
Matrine promotes NT3 expression in CNS cells in experimental autoimmune encephalomyelitis.
Neuroscience letters
Zhang ML,Zhang XJ,Kang J,Zhang HJ,Chen XL,Liu N,Liu SQ,Ma WD,Zhang GX,Zhu L
Wed May 10 00:00:00 EDT 2017
Matrine promotes NT3 expression in CNS cells in experimental autoimmune encephalomyelitis.
Neuroscience letters
Zhang ML,Zhang XJ,Kang J,Zhang HJ,Chen XL,Liu N,Liu SQ,Ma WD,Zhang GX,Zhu L
Wed May 10 00:00:00 EDT 2017
A(H1N1) vaccination recruits T lymphocytes to the choroid plexus for the promotion of hippocampal neurogenesis and working memory in pregnant mice.
Brain, behavior, and immunity
Qi F,Yang J,Xia Y,Yuan Q,Guo K,Zou J,Yao Z
Tue Mar 01 00:00:00 EST 2016
A(H1N1) vaccination recruits T lymphocytes to the choroid plexus for the promotion of hippocampal neurogenesis and working memory in pregnant mice.
Brain, behavior, and immunity
Qi F,Yang J,Xia Y,Yuan Q,Guo K,Zou J,Yao Z
Tue Mar 01 00:00:00 EST 2016
Autophagic flux regulates microglial phenotype according to the time of oxygen-glucose deprivation/reperfusion.
International immunopharmacology
Xia CY,Zhang S,Chu SF,Wang ZZ,Song XY,Zuo W,Gao Y,Yang PF,Chen NH
Sat Oct 01 00:00:00 EDT 2016
Autophagic flux regulates microglial phenotype according to the time of oxygen-glucose deprivation/reperfusion.
International immunopharmacology
Xia CY,Zhang S,Chu SF,Wang ZZ,Song XY,Zuo W,Gao Y,Yang PF,Chen NH
Sat Oct 01 00:00:00 EDT 2016
Conditioned Medium from the Stem Cells of Human Exfoliated Deciduous Teeth Ameliorates Experimental Autoimmune Encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Shimojima C,Takeuchi H,Jin S,Parajuli B,Hattori H,Suzumura A,Hibi H,Ueda M,Yamamoto A
Sun May 15 00:00:00 EDT 2016
Conditioned Medium from the Stem Cells of Human Exfoliated Deciduous Teeth Ameliorates Experimental Autoimmune Encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Shimojima C,Takeuchi H,Jin S,Parajuli B,Hattori H,Suzumura A,Hibi H,Ueda M,Yamamoto A
Sun May 15 00:00:00 EDT 2016
Neonatal BCG vaccination of mice improves neurogenesis and behavior in early life.
Brain research bulletin
Yang J,Qi F,Gu H,Zou J,Yang Y,Yuan Q,Yao Z
Fri Jan 01 00:00:00 EST 2016
Neonatal BCG vaccination of mice improves neurogenesis and behavior in early life.
Brain research bulletin
Yang J,Qi F,Gu H,Zou J,Yang Y,Yuan Q,Yao Z
Fri Jan 01 00:00:00 EST 2016
Effects of IL-4 on pulmonary fibrosis and the accumulation and phenotype of macrophage subpopulations following thoracic irradiation.
International journal of radiation biology
Groves AM,Johnston CJ,Misra RS,Williams JP,Finkelstein JN
Thu Dec 01 00:00:00 EST 2016
Effects of IL-4 on pulmonary fibrosis and the accumulation and phenotype of macrophage subpopulations following thoracic irradiation.
International journal of radiation biology
Groves AM,Johnston CJ,Misra RS,Williams JP,Finkelstein JN
Thu Dec 01 00:00:00 EST 2016
Pioglitazone suppresses neuronal and muscular degeneration caused by polyglutamine-expanded androgen receptors.
Human molecular genetics
Iida M,Katsuno M,Nakatsuji H,Adachi H,Kondo N,Miyazaki Y,Tohnai G,Ikenaka K,Watanabe H,Yamamoto M,Kishida K,Sobue G
Thu Jan 15 00:00:00 EST 2015
Pioglitazone suppresses neuronal and muscular degeneration caused by polyglutamine-expanded androgen receptors.
Human molecular genetics
Iida M,Katsuno M,Nakatsuji H,Adachi H,Kondo N,Miyazaki Y,Tohnai G,Ikenaka K,Watanabe H,Yamamoto M,Kishida K,Sobue G
Thu Jan 15 00:00:00 EST 2015
M2 Polarization of Monocytes-Macrophages Is a Hallmark of Indian Post Kala-Azar Dermal Leishmaniasis.
PLoS neglected tropical diseases
Mukhopadhyay D,Mukherjee S,Roy S,Dalton JE,Kundu S,Sarkar A,Das NK,Kaye PM,Chatterjee M
Thu Oct 01 00:00:00 EDT 2015
M2 Polarization of Monocytes-Macrophages Is a Hallmark of Indian Post Kala-Azar Dermal Leishmaniasis.
PLoS neglected tropical diseases
Mukhopadhyay D,Mukherjee S,Roy S,Dalton JE,Kundu S,Sarkar A,Das NK,Kaye PM,Chatterjee M
Thu Oct 01 00:00:00 EDT 2015
Ethanol supports macrophage recruitment and reinforces invasion and migration of Lewis lung carcinoma.
Alcoholism, clinical and experimental research
Yu K,Yang J,Wang F,Chen L,Lu Y,Luo J,Wang S
Wed Oct 01 00:00:00 EDT 2014
Ethanol supports macrophage recruitment and reinforces invasion and migration of Lewis lung carcinoma.
Alcoholism, clinical and experimental research
Yu K,Yang J,Wang F,Chen L,Lu Y,Luo J,Wang S
Wed Oct 01 00:00:00 EDT 2014
Ethanol supports macrophage recruitment and reinforces invasion and migration of Lewis lung carcinoma.
Alcoholism, clinical and experimental research
Yu K,Yang J,Wang F,Chen L,Lu Y,Luo J,Wang S
Wed Oct 01 00:00:00 EDT 2014
Ethanol supports macrophage recruitment and reinforces invasion and migration of Lewis lung carcinoma.
Alcoholism, clinical and experimental research
Yu K,Yang J,Wang F,Chen L,Lu Y,Luo J,Wang S
Wed Oct 01 00:00:00 EDT 2014