Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLRGSLSR LLQTRVHSIL KKSVHSVAVI GAPFSQGQKR KGVEHGPAAI REAGLMKRLS SLGCHLKDFG DLSFTPVPKD DLYNNLIVNP RSVGLANQEL AEVVSRAVSD GYSCVTLGGD HSLAIGTISG HARHCPDLCV VWVDAHADIN TPLTTSSGNL HGQPVSFLLR ELQDKVPQLP GFSWIKPCIS SASIVYIGLR DVDPPEHFIL KNYDIQYFSM RDIDRLGIQK VMERTFDLLI GKRQRPIHLS FDIDAFDPTL APATGTPVVG GLTYREGMYI AEEIHNTGLL SALDLVEVNP QLATSEEEAK TTANLAVDVI ASSFGQTREG GHIVYDQLPT PSSPDESENQ ARVRI (1-354 aa encoded by BC001350)
Arginase catalyzes the hydrolysis of arginine to ornithine and urea. At least two isoforms of mammalian arginase exists (types I and II) which differ in their tissue distribution, subcellular localization, immunologic crossreactivity and physiologic function. The type II isoform encoded by this gene, is located in the mitochondria and expressed in extra-hepatic tissues, especially kidney. The physiologic role of this isoform is poorly understood; it is thought to play a role in nitric oxide and polyamine metabolism. Transcript variants of the type II gene resulting from the use of alternative polyadenylation sites have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AII type II arginase; Arginase II; arginase type II; arginase, type II; Arginase-2, mitochondrial; kidney arginase; Kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; Non-hepatic arginase; nonhepatic arginase; Type II arginase
Gene Aliases: AII; ARG2; AU022422
UniProt ID: (Human) P78540, (Rat) O08701, (Mouse) O08691
Entrez Gene ID: (Human) 384, (Rat) 29215, (Mouse) 11847
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support