Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MTEKAPEPHV EEDDDDELDS KLNYKPPPQK SLKELQEMDK DDESLIKYKK TLLGDGPVVT DPKAPNVVVT RLTLVCESAP GPITMDLTGD LEALKKETIV LKEGSEYRVK IHFKVNRDIV SGLKYVQHTY RTGVKVDKAT FMVGSYGPRP EEYEFLTPVE EAPKGMLARG TYHNKSFFTD DDKQDHLSWE WNLSIKKEWT E
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D4; d4 gdi; GDI LYGDI; Ly-GDI; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP-dissociation inhibitor 2; Rho-GDI beta
Gene Aliases: ARHGDIB; D4; GDIA2; GDID4; Ly-GDI; LYGDI; RAP1GN1; RhoGDI2
UniProt ID: (Human) P52566, (Mouse) Q61599
Entrez Gene ID: (Human) 397, (Mouse) 11857
Molecular Function:
G-protein modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support