Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARH5; DKFZp686N1969; Ephexin 3; Ephexin-3; Guanine nucleotide regulatory protein TIM; Oncogene TIM; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; Transforming immortalized mammary oncogene
Gene Aliases: ARHGEF5; GEF5; P60; TIM; TIM1
UniProt ID: (Human) Q12774
Entrez Gene ID: (Human) 7984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support