Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGGFSRRCSK LINSSQLLYQ EYSDVVLNKE IQSQQRLESL SETPGPSSPR QPRKALVSSE SYLQRLSMAS SGSLWQEIPV VRNSTVLLSM THEDQKLQEV KFELIVSEAS YLRSLNIAVD HFQLSTSLRA TLSNQEHQWL FSRLQDVRDV SATFLSDLEE NFENNIFSFQ VCDVVLNHAP DFRRVYLPYV TNQTYQERTF QSLMNSNSNF REVLEKLESD PVCQRLSLKS FLILPFQRIT RLKLLLQNIL KRTQPGSSEE AEATKAHHAL EQLIRDCNNN VQSMRRTEEL IYLSQKIEFE CKIFPLISQS RWLVKSGELT ALEFSASPGL RRKLNTRPVH LHLFNDCLLL SRPREGSRFL VFDHAPFSSI RGEKCEMKLH GPHKNLFRLF LRQNTQGAQA EFLFRTETQS EKLRWISALA MPREELDLLE CYNSPQVQCL RAYKPRENDE LALEKADVVM VTQQSSDGWL EGVRLSDGER GWFPVQQVEF ISNPEVRAQN LKEAHRVKTA KLQLVEQQA
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARH5; DKFZp686N1969; Ephexin 3; Ephexin-3; Guanine nucleotide regulatory protein TIM; Oncogene TIM; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; Transforming immortalized mammary oncogene
Gene Aliases: ARHGEF5; GEF5; P60; TIM; TIM1
UniProt ID: (Human) Q12774
Entrez Gene ID: (Human) 7984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support