Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNPEEQIVTW LISLGVLESP KKTICDPEEF LKSSLKNGVV LCKLINRLMP GSVEKFCLDP QTEADCINNI NDFLKGCATL QVEIFDPDDL YSGVNFSKVL STLLAVNKAT EDQLSERPCG RSSSLSAANT SQTNPQGAVS STVSGLQRQS KTVEMTENGS HQLIVKARFN FKQTNEDELS VCKGDIIYVT RVEEGGWWEG TLNGRTGWFP SNYVREIKSS ERPLSPKAVK GFETAPLTKN YYTVVLQNIL DTEKEYAKEL QSLLVTYLRP LQSNNNLSTV EVTSLLGNFE EVCTFQQTLC QALEECSKFP ENQHKVGGCL LSLMPHFKSM YLAYCANHPS AVNVLTQHSD ELEQFMENQG ASSPGILILT TNLSKPFMRL EKYVTLLQEL ERHMEDTHPD HQDILKAIVA FKTLMGQCQD LRKRKQLELQ ILSEPIQAWE GEDIKNLGNV IFMSQVMVQY GACEEKEERY LMLFSNVLIM LSASPRMSGF IYQGKIPIAG TVVTRLDEIE GNDCTFEITG NTVERIVVHC NNNQDFQEWL EQLNRLIRGP ASCSSLSKTS SSSCSAHSSF SSTGQPRGPL EPPQIIKPWS LSCLRPAPPL RPSAALGYKE RMSYILKESS KSPKTMKKFL HKRKTERKPS EEEYVIRKST AALEEDAQIL KVIEAYCTSA NFQQGHGSST RKDSIPQVLL PEEEKLIIEE TRSNGQTIME EKSLVDTVYA LKDEVRELKQ ENKRMKQCLE EELKSRRDLE KLVRRLLKQT DECIRGESSS KTSILP
Acts as a RAC1 guanine nucleotide exchange factor (GEF).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-Pix; COOL-2; PAK-interacting exchange factor alpha; PAK-interacting exchange factor, alpha; Rac/Cdc42 guanine exchange factor (GEF) 6; Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6; Rac/Cdc42 guanine nucleotide exchange factor 6; Rho guanine nucleotide exchange factor 6
Gene Aliases: alpha-PIX; alphaPIX; ARHGEF6; Cool-2; COOL2; KIAA0006; MRX46; PIXA
UniProt ID: (Human) Q15052
Entrez Gene ID: (Human) 9459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support