Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLGNKRDLPN ALDEKQLIEK MNLSAIQDRE ICCYSISCKE KDNIDITLQW LIQHSKSRRS; Positive Samples: U-87MG, LO2, Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Cytoplasm, Late endosome membrane, Lysosome membrane, cytoskeleton, spindle
ARL8B may play a role in lysosome motility. may play a role in chromosome segregation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1
Gene Aliases: 2610313E07Rik; 3100002J04Rik; ARL10C; ARL8B; GIE1
UniProt ID: (Human) Q9NVJ2, (Mouse) Q9CQW2
Entrez Gene ID: (Human) 55207, (Mouse) 67166
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support