Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MATPAAVNPP EMASDIPGSV TLPVAPMAAT GQVRMAGAMP ARGGKRRSGM DFDDEDGEGP SKFSRENHSE IERRRRNKMT QYITELSDMV PTCSALARKP DKLTILRMAV SHMKSMRGTG NKSTDGAYKP SFLTEQELKH LILEAADGFL FVVAAETGRV IYVSDSVTPV LNQPQSEWFG STLYEQVHPD DVEKLREQLC TSENSMTGRI LDLKTGTVKK EGQQSSMRMC MGSRRSFICR MRCGNAPLDH LPLNRITTMR KRFRNGLGPV KEGEAQYAVV HCTGYIKAWP PAGMTIPEED ADVGQGSKYC LVAIGRLQVT SSPVCMDMNG MSVPTEFLSR HNSDGIITFV DPRCISVIGY QPQDLLGKDI LEFCHPEDQS HLRESFQQVV KLKGQVLSVM YRFRTKNREW MLIRTSSFTF QNPYSDEIEY IICTNTNVKQ LQQQQAELEV HQRDGLSSYD LSQVPVPNLP AGVHEAGKSV EKADAIFSQE RDPRFAEMFA GISASEKKMM SSASAAGTQQ IYSQGSPFPS GHSGKAFSSS VVHVPGVNDI QSSSSTGQNM SQISRQLNQS QVAWTGSRPP FPGQQIPSQS SKTQSSPFGI GTSHTYPADP SSYSPLSSPA TSSPSGNAYS SLANRTPGFA ESGQSSGQFQ GRPSEVWSQW QSQHHGQQSG EQHSHQQPGQ TEVFQDMLPM PGDPGNEWWS PHSRQHFRQP ISYARM
AhR, ARNT1, ARNT2 and BMAL1 are members of a family of transcription factors that contain a basic helix-loop-helix motif and a common "PAS" motif. The aromatic (aryl) hydrocarbon receptor, AhR, is a ligand dependent transcription factor that interacts with specific DNA sequences termed xenobiotic responsive elements (XREs) to activate several genes including CYP1A1, glutathione S-transferase Ya subunit and DT-diaphorase. The Ah receptor nuclear translocator proteins (Arnt 1 or Arnt 2) are required for ligand-dependent nuclear translocation of the Ah receptor and are also necessary for Ah receptor binding to the XRE element. BMAL1 (Brain and Muscle Arnt-like protein 1), also designated Arnt 3, TIC, JAP3 or MOP-3, has been shown to dimerize with Clock and bind to the promoter region of mPer1, suggesting that this protein plays a role in regulation of circadian oscillation in mammals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARNT 2; ARNT protein 2; Aryl hydrocarbon receptor nuclear translocator 2; aryl-hydrocarbon receptor nuclear translocator 2; bHLHe1; Class E basic helix-loop-helix protein 1
Gene Aliases: ARNT2; BHLHE1; KIAA0307; WEDAS
UniProt ID: (Human) Q9HBZ2
Entrez Gene ID: (Human) 9915
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support