Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARNTLSSR FRRVDIDEFD ENKFVDEQEE AAAAAAEPGP DPSEVDGLLR QGDMLRAFHA ALRNSPVNTK NQAVKERAQG VVLKVLTNFK SSEIEQAVQS LDRNGVDLLM KYIYKGFEKP TENSSAVLLQ WHEKALAVGG LGSIIRVLTA RKTV (1-153 aa encoded by BC000798)
May function as component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin related protein 2/3 complex subunit 5-like; Actin-related protein 2/3 complex subunit 5-like protein; ARC16-2; Arp2/3 complex 16 kDa subunit 2
Gene Aliases: 2010015J01Rik; AI852867; ARC16-2; ARPC5L; AW555592; AW742746
UniProt ID: (Human) Q9BPX5, (Rat) A1L108, (Mouse) Q9D898
Entrez Gene ID: (Human) 81873, (Rat) 296710, (Mouse) 74192
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support