Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alternative-splicing factor 1; ASF-1; ASF/SF2; pre-mRNA-splicing factor SF2, P33 subunit; Pre-mRNA-splicing factor SRp30a; Serine/arginine-rich splicing factor 1; splicing factor 2; Splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1
Gene Aliases: 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; ASF; AW491331; OK/SW-cl.3; SF2; SF2P33; SFRS1; SRp30a; SRSF1
UniProt ID: (Human) Q07955, (Mouse) Q5SXC5
Entrez Gene ID: (Human) 6426, (Rat) 689890, (Mouse) 110809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support