Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to the 23 kDa ASF1A protein.
Immunogen sequence: INWEDNTEK LEDAESSNPN LQSLLSTDAL PSASKGWSTS ENSLNVMLES HMDCM (151-204 aa encoded by BC010878)
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anti-silencing function protein 1 homolog A; ASF1 anti-silencing function 1 homolog A; CCG1-interacting factor A; CIA; DKFZP547E2110; hAsf1; hAsf1a; hCIA; Histone chaperone ASF1A; RP3-329L24.1
Gene Aliases: 2310079C17Rik; ASF1A; C79052; CGI-98; CIA; HSPC146
UniProt ID: (Human) Q9Y294, (Mouse) Q9CQE6
Entrez Gene ID: (Human) 25842, (Rat) 294408, (Mouse) 66403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support