Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTKEYQDLQ HLDNEESDHH QLRKGPPPPQ PLLQRLCSGP RLLLLSLGLS LLLLVVVCVI GSQNSQLQEE LRGLRETFSN FTASTEAQVK GLSTQGGNVG RKMKSLESQL EKQQKDLSED HSSLLLHVKQ FVSDLRSLSC QMAALQGNGS ERTCCPVNWV EHERSCYWFS RSGKAWADAD NYCRLEDAHL VVVTSWEEQK FVQHHIGPVN TWMGLHDQNG PWKWVDGTDY ETGFKNWRPE QPDDWYGHGL GGGEDCAHFT DDGRWNDDVC QRPYRWVCET ELDKASQEPP LL (1-291 aa encoded by BC032130)
The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalization and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i. e. it is present on the sinusoidal and lateral plasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed. The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASGP-R 1; ASGPR 1; Asialoglycoprotein receptor 1; C-type lectin domain family 4 member H1; Hepatic lectin 1; Hepatic lectin H1; HL-1
Gene Aliases: ASGPR; ASGPR1; Asgr; Asgr-1; ASGR1; CLEC4H1; HL-1
UniProt ID: (Human) P07306, (Mouse) P34927
Entrez Gene ID: (Pig) 100625649, (Human) 432, (Mouse) 11889
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support