Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: HGYPLEHPFN KDGYRYILAE PDPHAPDPEK LELDCWAGKP IPGDLYRACL YERVLLALHD RAPQLKISDD RLTVVGEKGY SMVRASHGVR KGAWYFEITV DEMPPDTAAR LGWSQPLGNL QAPLGYDKFS YSWRSKKGTK FHQSIGKHYS SGYGQGDVLG FYINLPEDTE TAKSLPDTYK DKALIKFKSY LYFEEKDFVD KAEKSLKQTP HSEIIFYKNG VNQGVAYKDI FEGVYFPAIS LYKSCTVSIN FGPCFKYPPK DLTYRPMSDM GWGAVVEHTL ADVLYHVETE VDGRRSPPWE P
The human ASH2L gene encodes a 628 amino acid protein known as ASH2L1, or isoform 1, which contains a nuclear localization signal and PHD finger motif, suggesting that the gene product functions as a transcription regulator. Its predicted molecular weight is 68.7kDa. Alternative splicing results in possible isoforms at 60 kDa & 56.5 kDa). Human ASH2L proteins are 60% homologous to Drosophila ash2, which positively regulates expression of certain genes in early development, and contain similar, but not identical, domains, including a zinc finger motif. Homology to mouse ASH2L is 90%. ASH2L is predominantly expressed in adult heart and testis and fetal lung and liver, with barely detectable expression in adult lung, liver, kidney, prostate, and peripheral leukocytes as well as leukemia cell lines with erythroid and megakaryocytic potential, such as K562, Hel and Dami. Differentiation inducers (e.g. phorbol ester and hemin) cause different expression patterns in these cell lines, suggesting that ASH2L plays a role in hematopoiesis and is associated with particular types of leukemia. In addition ASH2L has been suggested to interact with beta-tubulin and beta-catenin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASH 2; ash2 (absent, small, or homeotic)-like; ASH2 LIKE; ASH2 like protein; ASH2-like protein; Bre 2; Set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Aliases: ASH2; ASH2L; ASH2L1; ASH2L2; Bre2
UniProt ID: (Human) Q9UBL3
Entrez Gene ID: (Human) 9070, (Rat) 290829
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support