Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: NDQVVTDLRL WMRQTCSTLS GLLWELIRTM VDRAEAERDV LFPGYTHLQR AQPIRWSHWI LSHAVALTRD SERLLEVRKR INVLPLGSGA IAGNPLGVDR ELLRAELNFG AITLNSMDAT SERDFVAEFL FWASLCMTHL SRMAEDLILY CTKEFSFVQL SDAYSTGSSL MPQKKNPDSL ELIRSKAGRV FGRCAGLLMT LKGLPSTYNK DLQEDKEAVF EVSDTMSAVL QVATGVISTL QIHQENMGQA LSPDMLATDL AYYLVRKGMP FRQAHEASGK AVFMAETKGV ALNQLSLQEL QTISPLFSGD VICVWDYGHS VEQYGALGGT ARSSVDWQIR QVRALLQAQQ A
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: argininosuccinase; Argininosuccinate lyase; Arginosuccinase; ASAL
Gene Aliases: 2510006M18Rik; ASAL; ASL
UniProt ID: (Human) P04424, (Mouse) Q91YI0, (Rat) P20673
Entrez Gene ID: (Human) 435, (Pig) 100517757, (Mouse) 109900, (Rat) 59085
Molecular Function: lyase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support